Total Oxidant Status (TOS) Colorimetric Assay Kit
Invitrogen™

Total Oxidant Status (TOS) Colorimetric Assay Kit

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Have Questions?
Catalog NumberQuantity
EEA02796 tests
Catalog number EEA027
Price (INR)
42,392.00
線上優惠
Ends: 31-Dec-2026
70,653.00
Save 28,261.00 (40%)
Each
Quantity:
96 tests
Price (INR)
42,392.00
線上優惠
Ends: 31-Dec-2026
70,653.00
Save 28,261.00 (40%)
Each

The Total Oxidant Status (TOS) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of TOS in cells, tissue, serum, and other liquid sample types. Oxidant molecules, also known as reactive oxygen species (ROS), are produced during normal metabolic processes and can be further stimulated by internal and external factors. When the balance between oxidation and antioxidant defense shifts towards oxidation, it can lead to oxidative stress and various diseases. Measuring the concentrations of different oxidant molecules individually can be time-consuming, costly, and technically complex. Therefore, the Total Oxidant Status (TOS) assay provides a practical solution by quantifying the overall oxidation state of a sample.

In this assay, oxidizing agents in the sample oxidize ferrous ions (Fe2+) to ferric ions (Fe3+), which strongly bind to xylenol orange to produce a blue-purple complex with a maximum absorption wavelength of around 590 nm. The color depth is proportional to the content of oxidation substances in a certain concentration and a certain time and can be used to indirectly calculate the total oxidation state of the sample.

Key features

  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results
Specifications
Assay Range2.5 μmol H2O2 Equiv./L
Assay Sensitivity2.5 μmol H2O2 Equiv./L
For Use With (Equipment)Microplate Reader
Incubation Time5 min.
Interassay CV3.5
Intraassay CV2.3
Quantity96 tests
Sample Volume20 μL
Shipping ConditionDry Ice, 2–8°C
Target Kit NamedTotal Oxidant Status (TOS)
Total Assay Time30 min.
Detection MethodColorimetric
Sample TypeSerum, Tissue Homogenates
Unit SizeEach
Contents & Storage
Store at 2–8°C.