Free Fatty Acids (NEFA/FFA) Fluorometric Assay Kit
Invitrogen™

Free Fatty Acids (NEFA/FFA) Fluorometric Assay Kit

The Free Fatty Acids (NEFA/FFA) Fluorometric Assay Kit provides a sensitive and convenient method for quantifying free fatty acids in various samples.
Have Questions?
Catalog NumberQuantity
EEA03096 Tests
Catalog number EEA030
Price (INR)
41,667.00
Online offer
Ends: 30-Sep-2026
75,757.00
Save 34,090.00 (45%)
Each
Quantity:
96 Tests
Price (INR)
41,667.00
Online offer
Ends: 30-Sep-2026
75,757.00
Save 34,090.00 (45%)
Each

Free fatty acids are converted to acyl coenzyme A in the presence of acyl synthase. This acyl coenzyme A then reacts with acyl oxidase to produce hydrogen peroxide. In the presence of a specific enzyme and probe, hydrogen peroxide reacts to form a fluorescent substrate. The fluorescence intensity, measured at an excitation wavelength of 535 nm and an emission wavelength of 590 nm, is directly proportional to the concentration of free fatty acids in the sample. This kit is useful for researchers studying lipid metabolism, diabetes, obesity, and related fields.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
For Research Use Only.
Specifications
Assay Range1.20 to 100 μmol/L
Assay Sensitivity1.20 μmol/L
For Use With (Equipment)Fluorescence Microtiter Plate Reader
Incubation Time70 min.
Interassay CV5.1%
Intraassay CV4.1%
Product TypeKit
Quantity96 Tests
Sample Volume10 μL
Shipping ConditionDry Ice, 2°C to 8°C
Target Kit NamedFree Fatty Acids (NEFA/FFA)
Total Assay Time95 min.
Detection MethodFluorometric
Immunoassay Kit FormatFluorescent Detection Kit
Sample TypeSerum, Plasma, Animal Tissue, Cells
Target SpeciesUniversal
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.