Glucose (Glu) Colorimetric Assay Kit
Invitrogen™

Glucose (Glu) Colorimetric Assay Kit

With this kit, glucose oxidase catalyzes the oxidation of glucose to gluconic acid to produce hydrogen peroxide.
Have Questions?
Catalog NumberQuantity
EEA03296 Tests
Catalog number EEA032
Price (INR)
41,667.00
キャンペーン価格
Ends: 30-Sep-2026
75,757.00
Save 34,090.00 (45%)
Each
Quantity:
96 Tests
Price (INR)
41,667.00
キャンペーン価格
Ends: 30-Sep-2026
75,757.00
Save 34,090.00 (45%)
Each

With this kit, glucose oxidase catalyzes the oxidation of glucose to gluconic acid to produce hydrogen peroxide. In the presence of chromogenic oxygen receptors, peroxidase catalyzes hydrogen peroxide and oxidizes pigment sources to form colored substances. The OD value at 505 nm is then measured and glucose content calculated indirectly. This kit is useful for researchers studying glucose metabolism and diabetes.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
For Research Use Only.
Specifications
Assay Range0.04 to 30 mmol/L
Assay Sensitivity0.04 mmol/L
For Use With (Equipment)Microtiter Plate Reader
Incubation Time15 min.
Interassay CV2.3%
Intraassay CV1.9%
Product TypeKit
Quantity96 Tests
Sample Volume3 μL
Shipping ConditionDry Ice, 2°C to 8°C
Target Kit NamedGlucose (Glu)
Total Assay Time30 min.
Detection MethodColorimetric
Immunoassay Kit FormatColorimetric Detection Kit
Sample TypeSerum, Plasma, Tissue
Target SpeciesUniversal
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.