Zinc (Zn) Colorimetric Assay Kit
Invitrogen™

Zinc (Zn) Colorimetric Assay Kit

With this kit, the zinc ion in a sample reacts with 5-Br-PADAP to produce a colored complex.
Have Questions?
Catalog NumberQuantity
EEA03396 Tests
Catalog number EEA033
Price (INR)
41,667.00
線上優惠
Ends: 30-Sep-2026
75,757.00
Save 34,090.00 (45%)
Each
Quantity:
96 Tests
Price (INR)
41,667.00
線上優惠
Ends: 30-Sep-2026
75,757.00
Save 34,090.00 (45%)
Each

With this kit, the zinc ion in a sample reacts with 5-Br-PADAP to produce a colored complex. The depth of color is directly proportional to the concentration of zinc ion. Zinc ion content can be calculated by measuring the OD value at 560 nm. This kit is useful for researchers studying zinc metabolism and its biological functions.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
For Research Use Only.
Specifications
Assay Range0.748 to 46.2 μmol/L
Assay Sensitivity0.418 μmol/L
For Use With (Equipment)Microtiter Plate Reader
Incubation Time5 min.
Interassay CV4.0%
Intraassay CV2.7%
Product TypeKit
Quantity96 Tests
Sample Volume50 μL
Shipping ConditionDry Ice, 2°C to 8°C
Target Kit NamedZinc (Zn)
Total Assay Time30 min.
Detection MethodColorimetric
Immunoassay Kit FormatColorimetric Detection Kit
Sample TypeSerum, Plasma, Urine, Milk
Target SpeciesUniversal
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.