Oxalate (Oxalic Acid) Colorimetric Assay Kit
Invitrogen™

Oxalate (Oxalic Acid) Colorimetric Assay Kit

With this kit, oxalate oxidase catalyzes the oxidation of oxalate to produce hydrogen peroxide and carbon dioxide.
Have Questions?
Catalog NumberQuantity
EEA03696 Tests
Catalog number EEA036
Price (INR)
41,667.00
Online offer
Ends: 30-Sep-2026
75,757.00
Save 34,090.00 (45%)
Each
Quantity:
96 Tests
Price (INR)
41,667.00
Online offer
Ends: 30-Sep-2026
75,757.00
Save 34,090.00 (45%)
Each

With this kit, oxalate oxidase catalyzes the oxidation of oxalate to produce hydrogen peroxide and carbon dioxide. Under the action of POD, hydrogen peroxide reacts with chromogenic substances to produce colored products. There is a specific absorption peak at 550 nm, and the color depth is proportional to the content of oxalate. This kit is useful for researchers studying oxalate metabolism and kidney function.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
For Research Use Only.
Specifications
Assay Range0.02 to 1 mmol/L
Assay Sensitivity0.02 mmol/L
For Use With (Equipment)Microtiter Plate Reader
Incubation Time10 min.
Interassay CV4.7%
Intraassay CV3.0%
Product TypeKit
Quantity96 Tests
Sample Volume10 μL
Shipping ConditionDry Ice, 2°C to 8°C
Target Kit NamedOxalate (Oxalic Acid)
Total Assay Time30 min.
Detection MethodColorimetric
Immunoassay Kit FormatColorimetric Detection Kit
Sample TypeSerum, Plasma, Plant Tissue
Target SpeciesUniversal
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.