Sodium (Na) Colorimetric Assay Kit
Invitrogen™

Sodium (Na) Colorimetric Assay Kit

With this kit, β-galactosidase activated by sodium ions catalyzes glycoside analogues to produce glycoside and o-nitrophenol.
Have Questions?
Catalog NumberQuantity
EEA03896 Tests
Catalog number EEA038
Price (INR)
75,757.00
Each
Quantity:
96 Tests
Price (INR)
75,757.00
Each

With this kit, β-galactosidase activated by sodium ions catalyzes glycoside analogues to produce glycoside and o-nitrophenol. The increase in absorbance at 405 nm is determined, and the content of sodium ion is calculated indirectly. This kit is useful for researchers studying sodium ion concentration and electrolyte balance.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
For Research Use Only.
Specifications
Assay Range0.02 to 20 mmol/L
Assay Sensitivity0.02 mmol/L
For Use With (Equipment)Microtiter Plate Reader
Incubation Time3 min.
Interassay CV8.4%
Intraassay CV2.2%
Product TypeKit
Quantity96 Tests
Sample Volume10 μL
Shipping ConditionDry Ice, 2°C to 8°C
Target Kit NamedSodium (Na)
Total Assay Time20 min.
Detection MethodColorimetric
Immunoassay Kit FormatColorimetric Detection Kit
Sample TypeSerum, Plasma, Tissue
Target SpeciesUniversal
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.