Lipoxygenase (LOX) Activity Assay Kit
Invitrogen™

Lipoxygenase (LOX) Activity Assay Kit

Lipoxygenase (LOX) is widely found in plant tissues, especially in seeds with high oil content such as soybean. LOX catalyzes the oxidation of unsaturated fatty acids, leading to membrane lipid peroxidation. It plays an important role in plant growth and development, maturation and senescence, and stress.
Have Questions?
Catalog NumberQuantity
EEA03996 Tests
Catalog number EEA039
Price (INR)
41,667.00
Online offer
Ends: 30-Sep-2026
75,757.00
Save 34,090.00 (45%)
Each
Quantity:
96 Tests
Price (INR)
41,667.00
Online offer
Ends: 30-Sep-2026
75,757.00
Save 34,090.00 (45%)
Each

With this kit, LOX catalyzes linoleic acid oxidation, and the oxidation product has a specific absorption peak at 280 nm. The activity of LOX can be calculated by measuring the change in absorbance value at 280 nm. This kit is useful for researchers studying plant physiology and lipid oxidation.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
For Research Use Only.
Specifications
Assay Range5 to 339.5 U/L
Assay Sensitivity5 U/L
For Use With (Equipment)Microtiter Plate Reader
Incubation Time1 min.
Interassay CV5.0%
Intraassay CV4.0%
Product TypeKit
Quantity96 Tests
Sample Volume20 μL
Shipping ConditionDry Ice, 2°C to 8°C
Target Kit NamedLipoxygenase (LOX)
Total Assay Time20 min.
Detection MethodColorimetric
Immunoassay Kit FormatColorimetric Detection Kit
Sample TypePlant Tissue
Target SpeciesUniversal
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.