Vitamin E (VE) Colorimetric Assay Kit
Invitrogen™

Vitamin E (VE) Colorimetric Assay Kit

With this kit, Fe3+ is deoxidized to Fe2+ by vitamin E.
Have Questions?
Catalog NumberQuantity
EEA04196 Tests
Catalog number EEA041
Price (INR)
75,757.00
Each
Quantity:
96 Tests
Price (INR)
75,757.00
Each

With this kit, Fe3+ is deoxidized to Fe2+ by vitamin E. The Fe2+ then reacts with phenanthroline to form a pink compound under certain conditions. Vitamin E content can be calculated by measuring the OD value at 532 nm. This kit is useful for researchers studying antioxidant activity and vitamin E metabolism.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
For Research Use Only.
Specifications
Assay Range0.09 to 40 μg/mL
Assay Sensitivity0.09 μg/mL
For Use With (Equipment)spectrophotometer
Incubation Time7 min.
Interassay CV4.9%
Intraassay CV4.4%
Product TypeKit
Quantity96 Tests
Sample Volume300 μL
Shipping ConditionDry Ice, 2°C to 8°C
Target Kit NamedVitamin E (VE)
Total Assay Time60 min.
Detection MethodColorimetric
Immunoassay Kit FormatColorimetric Detection Kit
Sample TypeSerum, Plasma, Tissue
Target SpeciesUniversal
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.