α-Amylase Activity Assay Kit
Invitrogen™

α-Amylase Activity Assay Kit

With this kit, a reducing sugar reacts with 3,5-dinitrosalicylic acid under heating conditions to produce a brown-red substance, which is inactivated by the thermolabile nature of β-amylase.
Have Questions?
Catalog NumberQuantity
EEA04496 Tests
Catalog number EEA044
Price (INR)
41,667.00
キャンペーン価格
Ends: 30-Sep-2026
75,757.00
Save 34,090.00 (45%)
Each
Quantity:
96 Tests
Price (INR)
41,667.00
キャンペーン価格
Ends: 30-Sep-2026
75,757.00
Save 34,090.00 (45%)
Each

With this kit, a reducing sugar reacts with 3,5-dinitrosalicylic acid under heating conditions to produce a brown-red substance, which is inactivated by the thermolabile nature of β-amylase. The enzyme activity of α-amylase is then determined. This kit is useful for researchers studying carbohydrate metabolism and enzyme activity.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
For Research Use Only.
Specifications
Assay Range0.0097 to 0.3474 U/mL
Assay Sensitivity0.0097 U/mL
For Use With (Equipment)Microtiter Plate Reader
Incubation Time10 min.
Interassay CV3.9%
Intraassay CV3.0%
Product TypeKit
Quantity96 Tests
Sample Volume75 μL
Shipping ConditionDry Ice, 2°C to 8°C
Target Kit Namedα-Amylase
Total Assay Time40 min.
Detection MethodColorimetric
Immunoassay Kit FormatColorimetric Detection Kit
Sample TypeSerum, Saliva, Animal Tissue, Plant Tissue
Target SpeciesUniversal
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.