Cysteine (Cys) Colorimetric Assay Kit
Invitrogen™

Cysteine (Cys) Colorimetric Assay Kit

With this kit, phosphotungstic acid is reduced by cysteine to form tungsten blue, which has an absorption peak at 600 nm.
Have Questions?
Catalog NumberQuantity
EEA04596 Tests
Catalog number EEA045
Price (INR)
41,667.00
온라인 행사
Ends: 30-Sep-2026
75,757.00
Save 34,090.00 (45%)
Each
Quantity:
96 Tests
Price (INR)
41,667.00
온라인 행사
Ends: 30-Sep-2026
75,757.00
Save 34,090.00 (45%)
Each

With this kit, phosphotungstic acid is reduced by cysteine to form tungsten blue, which has an absorption peak at 600 nm. Cysteine content can be calculated with the absorbance at 600 nm. This kit is useful for researchers studying amino acid metabolism and redox biology.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
For Research Use Only.
Specifications
Assay Range0.07 to 2.0 mmol/L
Assay Sensitivity0.03 mmol/L
For Use With (Equipment)Microtiter Plate Reader
Incubation Time10 min.
Interassay CV1.3%
Intraassay CV1.1%
Product TypeKit
Quantity96 Tests
Sample Volume20 μL
Shipping ConditionDry Ice, 2°C to 8°C
Target Kit NamedCysteine (Cys)
Total Assay Time30 min.
Detection MethodColorimetric
Immunoassay Kit FormatColorimetric Detection Kit
Sample TypeSerum, Plasma, Animal Tissue, Cells
Target SpeciesUniversal
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.