Trehalose Colorimetric Assay Kit
Invitrogen™

Trehalose Colorimetric Assay Kit

With this kit, trehalase hydrolyzes trehalose into two molecules of glucose, and the glucose content is detected by the GOD-POD method.
Have Questions?
Catalog NumberQuantity
EEA04896 Tests
Catalog number EEA048
Price (INR)
75,757.00
Each
Quantity:
96 Tests
Price (INR)
75,757.00
Each

Trehalose is found in a large number of organisms, including bacteria, algae, yeast, plants, insects, and other invertebrates. Because trehalose has unique biological characteristics different from other carbohydrates, it can protect the protein, lipid, carbohydrate, nucleic acid, and other components of biological cells from damage under harsh environments such as drought, high temperature, dehydration, freezing, high osmotic pressure, and toxic substances.

With this kit, trehalase hydrolyzes trehalose into two molecules of glucose, and the glucose content is detected by the GOD-POD method. Other disaccharides such as maltose and lactose do not interfere with the determination. This kit is useful for researchers studying carbohydrate metabolism and stress responses.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
For Research Use Only.
Specifications
Assay Range0.05 to 1.9 mg/mL
Assay Sensitivity0.05 mg/mL
For Use With (Equipment)Microtiter Plate Reader
Incubation Time30 min.
Interassay CV4.8%
Intraassay CV3.2%
Product TypeKit
Quantity96 Tests
Sample Volume10 μL
Shipping ConditionDry Ice, 2°C to 8°C
Target Kit NamedTrehalose
Total Assay Time60 min.
Detection MethodColorimetric
Immunoassay Kit FormatColorimetric Detection Kit
Sample TypeSerum, Plasma, Tissue, Cells
Target SpeciesUniversal
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.