Proline (Pro) Colorimetric Assay Kit
Invitrogen™

Proline (Pro) Colorimetric Assay Kit

With this kit, proline reacts with acidic ninhydrin to form a stable red compound.
Have Questions?
Catalog NumberQuantity
EEA051100 Tests
Catalog number EEA051
Price (INR)
41,667.00
Online offer
Ends: 30-Sep-2026
75,757.00
Save 34,090.00 (45%)
Each
Quantity:
100 Tests
Price (INR)
41,667.00
Online offer
Ends: 30-Sep-2026
75,757.00
Save 34,090.00 (45%)
Each

With this kit, proline reacts with acidic ninhydrin to form a stable red compound. The maximum absorption peak of the compound is at 520 nm. The concentration of proline can be calculated by measuring the OD value at 520 nm. This kit is useful for researchers studying amino acid metabolism and stress responses.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
For Research Use Only.
Specifications
Assay Range0.17 to 35 mmol/L
Assay Sensitivity0.17 mmol/L
For Use With (Equipment)spectrophotometer
Incubation Time0 min.
Interassay CV5.4%
Intraassay CV4.5%
Product TypeKit
Quantity100 Tests
Sample Volume2 mL
Shipping ConditionDry Ice, 2°C to 8°C
Target Kit NamedProline (Pro)
Total Assay Time60 min.
Detection MethodColorimetric
Immunoassay Kit FormatColorimetric Detection Kit
Sample TypePlant Tissue, Honey
Target SpeciesUniversal
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.