Lipase (LPS) Activity Assay Kit
Invitrogen™

Lipase (LPS) Activity Assay Kit

With this kit, lipase catalyzes a substrate to produce sulfhydryl compounds, which react with DTNB to generate TNB.
Have Questions?
Catalog NumberQuantity
EEA05296 Tests
Catalog number EEA052
Price (INR)
41,667.00
線上優惠
Ends: 30-Sep-2026
75,757.00
Save 34,090.00 (45%)
Each
Quantity:
96 Tests
Price (INR)
41,667.00
線上優惠
Ends: 30-Sep-2026
75,757.00
Save 34,090.00 (45%)
Each

With this kit, lipase catalyzes a substrate to produce sulfhydryl compounds, which react with DTNB to generate TNB. TNB has a maximum absorption at 412 nm. The activity of lipase in the sample can be calculated by measuring the change of absorbance per unit time. This kit is useful for researchers studying lipid metabolism and enzyme activity.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
For Research Use Only.
Specifications
Assay Range0.03 to 9.41 U/L
Assay Sensitivity0.03 U/L
For Use With (Equipment)Microplate Reader
Incubation Time30 min.
Interassay CV6.0%
Intraassay CV3.0%
Product TypeKit
Quantity96 Tests
Sample Volume10 μL
Shipping ConditionDry Ice, 2°C to 8°C
Target Kit NamedLipase (LPS)
Total Assay Time60 min.
Detection MethodColorimetric
Immunoassay Kit FormatColorimetric Detection Kit
Sample TypeSerum, Plasma, Animal Tissue, Cells
Target SpeciesUniversal
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.