Plant Flavonoids Colorimetric Assay Kit
Invitrogen™

Plant Flavonoids Colorimetric Assay Kit

With this kit, in alkaline nitrite solution, flavonoids form a red complex with aluminum ion.
Have Questions?
Catalog NumberQuantity
EEA05796 Tests
Catalog number EEA057
Price (INR)
41,667.00
Online offer
Ends: 30-Sep-2026
75,757.00
Save 34,090.00 (45%)
Each
Quantity:
96 Tests
Price (INR)
41,667.00
Online offer
Ends: 30-Sep-2026
75,757.00
Save 34,090.00 (45%)
Each

With this kit, in alkaline nitrite solution, flavonoids form a red complex with aluminum ion. The flavonoid content of the sample can be calculated by measuring the absorbance of the sample extract at 510 nm. This kit is useful for researchers studying plant secondary metabolites and antioxidant activity.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
For Research Use Only.
Specifications
Assay Range0.315 to 150 μg/mL
Assay Sensitivity0.315 μg/mL
For Use With (Equipment)Spectrophotometer
Incubation Time15 min.
Interassay CV2.2%
Intraassay CV1.9%
Product TypeKit
Quantity96 Tests
Sample Volume0.54 mL
Shipping ConditionDry Ice, 2°C to 8°C
Target Kit NamedPlant Flavonoids
Total Assay Time30 min.
Detection MethodColorimetric
Immunoassay Kit FormatColorimetric Detection Kit
Sample TypePlant Tissue
Target SpeciesUniversal
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.