Replacement Parts for Electrophoresis ApparatusRead more
Have Questions?
Catalog Number
Quantity
EI0012
1 Each
Catalog number EI0012
Price (INR)
5,452.00
Each
Quantity:
1 Each
Price (INR)
5,452.00
Each
Replacement Parts for Electrophoresis Apparatus
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 Each
Shipping ConditionRoom Temperature
For Use With (Equipment)XCell SureLock™ Mini
Gel SizeMini
Product LineXCell II, XCell SureLock
TypeBuffer Dam
Unit SizeEach
Contents & Storage
Store at room temperature.
Frequently asked questions (FAQs)
Can I purchase replacement parts for the XCell SureLock Mini-Cell?
Here are the replacement parts we offer for the XCell SureLock Mini-Cell:
Replacement part - Cat. No.
Buffer Dam for XCell SureLock Mini-Cell and XCell II Blot Module - EI0012
L-Buffer Chamber for XCell SureLock Mini-Cell and XCell II Mini-Cell - EI0013
Platinum Wire for XCell SureLock Mini-Cell (12 inch x 2) - EI9022
XCell SureLock Lid - EI0010
XCell SureLock Wedge - EI0011
Buffer Core with Cathode Bar for XCell SureLock Mini-Cell - EI9015
Gel Knife - EI9010
Sponge Pad for Blotting (8) - EI9052