Buffer Core Repair Kit for XCell SureLock™ Mini-Cell
The Buffer Core Repair Kit for the XCell SureLock™ Mini-Cell includes a platinum wire, wire insulation tubing, banana plug, hexRead more
Have Questions?
Catalog Number
Quantity
EI9021
1 kit
Catalog number EI9021
Price (INR)
8,697.00
Each
Quantity:
1 kit
Price (INR)
8,697.00
Each
The Buffer Core Repair Kit for the XCell SureLock™ Mini-Cell includes a platinum wire, wire insulation tubing, banana plug, hex nut, and washer.
More About the XCell SureLock™ Mini-Cell The XCell SureLock™ Mini-Cell holds gels firmly with a simple gel tension wedge. When the lever on the gel tension wedge is pushed forward into the locked position, an even horizontal force is generated. This seals the gel/buffer core assembly firmly into position in the lower buffer chamber. The positive locking action of the gel tension wedge ensures a trouble-free, leak-free gel run every time.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 kit
Shipping ConditionRoom Temperature
For Use With (Application)1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
For Use With (Equipment)XCell SureLock™ Mini
Gel SizeMini
Product LineXCell SureLock
TypeMini-Cell Buffer Core Repair Kit
Unit SizeEach
Contents & Storage
Store at room temperature.
Frequently asked questions (FAQs)
Can I purchase replacement parts for the XCell SureLock Mini-Cell?
Here are the replacement parts we offer for the XCell SureLock Mini-Cell:
Replacement part - Cat. No.
Buffer Dam for XCell SureLock Mini-Cell and XCell II Blot Module - EI0012
L-Buffer Chamber for XCell SureLock Mini-Cell and XCell II Mini-Cell - EI0013
Platinum Wire for XCell SureLock Mini-Cell (12 inch x 2) - EI9022
XCell SureLock Lid - EI0010
XCell SureLock Wedge - EI0011
Buffer Core with Cathode Bar for XCell SureLock Mini-Cell - EI9015
Gel Knife - EI9010
Sponge Pad for Blotting (8) - EI9052