<i>Thermus thermophilus</i> DNA Ligase
Thermo Scientific™

Thermus thermophilus DNA Ligase

Thermo Scientific™ Thermus thermophilus DNA Ligase is a thermostable NAD-dependent DNA ligase that catalizes formation of phosphodiester bonds between 5'-phospate and 3'-hydroxyl termini in double-stranded DNA.
Have Questions?
Catalog NumberQuantity
EL079811000 U
Catalog number EL07981
Price (INR)
26,991.00
Online offer
Ends: 31-Dec-2026
44,985.00
Save 17,994.00 (40%)
Each
Quantity:
1000 U
Price (INR)
26,991.00
Online offer
Ends: 31-Dec-2026
44,985.00
Save 17,994.00 (40%)
Each

Thermo Scientific™ Thermus thermophilus DNA Ligase is a thermostable NAD-dependent DNA ligase that catalizes formation of phosphodiester bonds between 5`-phospate and 3`-hydroxyl termini in double-stranded DNA. Thermus thermophilus DNA Ligase is not active on RNA, single-stranded DNA, or blunt-ended DNA.

Features

  • Increased specificity—high ligation temperature eliminates non-specific ligation
  • Stabile at high temperatures with an optimum ligation temperature of 45°C to 65°C

Applications

Thermus thermophilus DNA ligase is recommended for applications requiring high ligation specificity, including:

  • Single-stranded nick repair in double-stranded DNA
  • Next-generation sequencing
  • Rolling circle amplification
  • Proximity ligation assay
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Single-stranded nick repair, NGS, RCA, PLA
Product TypeNAD-dependent DNA Ligase
Quantity1000 U
Concentration50 U/μL
EnzymeDNA Ligase
FormLiquid
Research CategoryNext-generation Sequencing, DNA Repair
Sensitive to Heat InactivationNo
Unit SizeEach
Contents & Storage
10X Thermus thermophilus DNA Ligase Buffer, 40 μL

Store at -20°C.