TheraPure™ 2'-O-Methyltransferase (125 U/μL)
Thermo Scientific™

TheraPure™ 2'-O-Methyltransferase (125 U/μL)

2'-O-Methyltransferase provides a precise and efficient means of converting Cap-0 to Cap-1 structures by adding a methyl group to the 2'-O position of the ribose sugar, further enhancing mRNA stability and translation efficiency. This enzyme is designed to work seamlessly with other capping reagents, ensuring a streamlined process that maximizes mRNA quality and functionality
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
EM16712TPB0110.5 mL
EM16712TPB0121 mL
EM16712TPB0088 mL
EM16712TPB00124 mL
Catalog number EM16712TPB011
Price (INR)
-
Quantity:
0.5 mL

Thermo Scientific TheraPure products for post-transcriptional capping are intended for use in the synthesis of mRNA in the development of mRNA therapeutics and vaccines.

Post-transcriptional capping involves adding a 5' cap structure to mRNA after transcription, enhancing mRNA stability and translation efficiency. The process uses TheraPure Vaccinia Capping Enzyme and TheraPure 2'-O-Methyltransferase for adding the cap structures and TheraPure S-Adenosyl-L-Methionine as a methyl donor. Capping efficiency can reach >95%.

Designed for process development

TheraPure Vaccinia Capping Enzyme is a highly efficient enzyme used for adding 7-methylguanylate cap structures (Cap-0) to the 5' ends of uncapped RNA. This capping is essential for the stability and translational efficiency of mRNA.

TheraPure 2'-O-Methyltransferase is a high-purity enzyme designed for the methylation of RNA molecules, specifically adding a methyl group to the 2'-O position of the ribose sugar (Cap-1).

TheraPure S-Adenosyl-L-Methionine is a high-purity reagent used as a methyl group donor in post-transcriptional capping for Cap-0 and Cap-1 structure.

These TheraPure products are essential tools in the field of molecular biology, providing researchers with reliable and high-quality enzymes and reagents to advance the development of mRNA therapeutics and vaccines.

For Research Use Only.
Specifications
Compatible Buffer10X Capping buffer
Product TypeRNA Methylation
Quantity0.5 mL
Shipping Condition-25°C to -15°C
Concentration125 U/μL
Enzyme2'-O-Methyltransferase
FormLiquid
Research CategoryDrug Development
Sensitive to Heat InactivationNo
Unit SizeEach