TheraPure™ GMP DNase I, RNase-free, 50 U/μL
Thermo Scientific™

TheraPure™ GMP DNase I, RNase-free, 50 U/μL

Thermo Scientific TheraPure GMP* DNase I, RNase-free, is manufactured to ICH Q7 GMP principles and supported by a comprehensive documentationRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
EN052SKB01250 kU
EN052R30SKB001250 kU
EN052SKB0082500 kU
EN052R3AFB01316000 kU
Catalog number EN052SKB012
Price (INR)
-
Quantity:
50 kU

Thermo Scientific TheraPure GMP* DNase I, RNase-free, is manufactured to ICH Q7 GMP principles and supported by a comprehensive documentation package. It is designed for DNA template removal during the development and manufacturing of mRNA therapeutics and vaccines. As an endonuclease it digests both single- and double-stranded DNA.

Designed for process development and manufacturing
TheraPure GMP DNase I, RNase-free helps accelerate mRNA vaccine and therapeutics production from preclinical development to commercialization by providing:
Quality—meet critical product quality needs to help accelerate therapeutic development and manufacturing
Minimal risk—helps reduce risks in developing therapeutics by utilizing proven products
Technical partnership—partner with expert product and technical support
Secure and consistent supply—count on a secure and stable supply of product at a global scale

Comprehensive quality system
TheraPure GMP DNase I is supported by a extensive quality system and documentation including:
• Manufactured to relevant ICH Q7 GMP guidelines
• Animal-origin free (AOF) manufacturing processes, materials, and facilities
• Validated manufacturing processes and analytical test methods
• Comprehensive impurity profiles
• Verified compendial assays where applicable
• Product stability data
• Tested by analytical methods following ICH Q2 guidelines whenever possible
• Drug master file (DMF)

Enzyme activity
One unit of DNase I completely degrades 1 μg of plasmid DNA in 10 minutes at 37°C. One DNase I unit is equivalent to 0.3 Kunitz unit.

For additional TheraPure GMP products or customization, please visit www.thermofisher.com/therapure.

*TheraPure GMP refers to the quality level of the raw, ancillary, or starting materials to be used for further manufacturing. TheraPure GMP products are manufactured in facilities with ISO 9001–certified quality management systems that operate in accordance with relevant good manufacturing practice (GMP) principles, as outlined in ICH Q7 or equivalent guidance documents or standards.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Biological ActivityDNase
ColorTransparent
Expression SystemPichia pastoris
For Use With (Application)In vitro transcription, DNA template removal
Mass29 kDa
Product TypeDNase
Purity≥95%
Quantity50 kU
SourceBovine gene
Storage Buffer50 mM Tris-HCl, pH 7.5, 10 mM CaCl2, 50% glycerol
Concentration50 U/μL
EnzymeDNase
FormLiquid
Product LineTheraPure™ GMP
pH7.1 to 7.9
Unit SizeEach