T3 RNA Polymerase, HC (≥100 U/μL)
Thermo Scientific™

T3 RNA Polymerase, HC (≥100 U/μL)

Thermo Scientific Bacteriophage T3 RNA polymerase is a DNA-dependent RNA polymerase with strict specificity for its respective double-stranded promoters. ItRead more
Have Questions?
Catalog NumberQuantity
EP010310,000 units
Catalog number EP0103
Price (INR)
-
Quantity:
10,000 units
Request bulk or custom format
Thermo Scientific Bacteriophage T3 RNA polymerase is a DNA-dependent RNA polymerase with strict specificity for its respective double-stranded promoters. It catalyzes the 5'→3' synthesis of RNA on either single-stranded DNA or double-stranded DNA downstream from it promoter and is able to incorporate modified nucleotide.

Highlights

• Incorporates modified nucleotides (e.g., aminoallyl-, biotin-, fluorescein-, digoxigenin-labeled nucleotides)

Applications

• Synthesis of unlabeled and labeled RNA that can be used:
• For hybridization, in vitro RNA translation
• As aRNA, siRNA, substrate in RNase protection assays, template for genomic DNA sequencing
• In studies of RNA secondary structure and RNA-protein interactions, RNA splicing

Note

Consensus promoter sequence:
T3: AATTAACCCTCACTAAAGGGAGA
The position in bold (+1) indicates the first nucleotide incorporated into RNA during transcription. Only bases at this position through +3 are critical for transcription, and they must be a G and a purine base, respectively.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration200U/µL
Quantity10,000 units
PolymeraseT3 RNA Polymerase
Unit SizeEach