TheraPure™ SuperFi™ II DNA Polymerase
Invitrogen™

TheraPure™ SuperFi™ II DNA Polymerase

TheraPure SuperFi II DNA Polymerase is a high-fidelity, proofreading DNA polymerase that combines exceptional accuracy with an optimized buffer system enabling a universal primer annealing temperature. Manufactured using animal-origin-free (AOF) and beta-lactam-free processes, it supports nucleic acid-based therapeutic development workflows.
Have Questions?
Catalog NumberQuantity
EP250TPB001200 Units
Catalog number EP250TPB001
Price (INR)
30,762.00
Each
Quantity:
200 Units
Price (INR)
30,762.00
Each

TheraPure SuperFi II DNA Polymerase is a high-fidelity, proofreading DNA polymerase that combines exceptional accuracy with an optimized buffer system enabling a universal primer annealing temperature. Manufactured using animal-origin-free (AOF) and beta-lactam-free processes, it supports nucleic acid-based therapeutic development workflows.

Key Features:

  • Animal-origin-free and beta-lactam-free manufacturing: Produced using processes and materials free from animal-derived components and beta-lactam compounds, supporting sensitive applications
  • exceptional fidelity performance: Delivers >300-fold higher fidelity than standard Taq polymerase for accurate DNA amplification
  • Extended amplification range: Enables long-range PCR of DNA targets up to 20 kb, suitable for large genomic regions and complex templates
  • Standardized primer annealing temperature: Universal 60°C annealing temperature simplifies PCR optimization and enables consistent thermal cycling across primer sets

TheraPure SuperFi II DNA Polymerase incorporates Affibody-based hot-start technology to inhibit enzyme activity at ambient temperatures, allowing room-temperature reaction setup and extended stability of pre-assembled PCR reactions. Enzyme activity is restored during the initial denaturation step enabling high specificity.

TheraPure™ 5X SuperFi™ II Buffer contains specialized additives that support efficient amplification of both AT-rich and GC-rich templates. Robust performance is achieved with GC-rich sequences up to 75% GC without the need for additional melting agents. Buffer additives stabilize primer–template duplexes during annealing, enabling increased specificity without individual primer optimization.

TheraPure SuperFi II DNA Polymerase possesses 5'→3' polymerase and 3'→5' exonuclease (proofreading) activities and lacks 5'→3' exonuclease activity. The enzyme generates blunt-end DNA products.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)>300X
Hot StartBuilt-In Hot Start
No. of Reactions100 Reactions
OverhangBlunt
Product TypeHigh-Fidelity DNA Polymerase
Quantity200 Units
Reaction FormatStandalone
Shipping ConditionFrozen Gel Pack
Concentration2 U/μL
Detection MethodGel Electrophoresis
For Use With (Application)Hot-start PCR, High-fidelity PCR
FormLiquid
GC-Rich PCR PerformanceHigh
PCR MethodStandard PCR, End-point PCR
Reaction SpeedFast
Unit SizeEach
Contents & Storage
TheraPure™ SuperFi™ II DNA Polymerase (100 μL); TheraPure™ 5X SuperFi™ II Buffer (2 x 1.25 mL)
Store at -20°C