ProcartaPlex™ Wash Buffer (10X)
Invitrogen™

ProcartaPlex™ Wash Buffer (10X)

ProcartaPlex Wash Buffer is for use with ProcartaPlex assays. It is supplied as a 10X concentrate and used for washing the ProcartaPlex assays using flat bottom plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
EPX-66666-001200 mL
EPX-66666-00025 mL
Catalog number EPX-66666-001
Price (INR)
-
Quantity:
200 mL

ProcartaPlex Wash Buffer is for use with ProcartaPlex assays. It is supplied as a 10X concentrate and used for washing the ProcartaPlex assays using flat bottom plates. It is a simple mix of buffers, salts, and mild detergent.

ProcartaPlex Wash Buffer (10X) is included in our basic kits and off-the-shelf panels. Additionally, we offer it here as a stand-alone item.

Reported application

  • Multiplex immunoassay
For Research Use Only.
Specifications
For Use With (Equipment)Luminex™ Instruments
Product TypeBuffer
Quantity200 mL
Unit SizeEach

Frequently asked questions (FAQs)

Are the standalone ProcartaPlex assay buffers lot-specific such that they will only work with specific lots of other components?

No, the standalone ProcartaPlex assay buffers are not lot-specific.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

At what temperature and for how long can the diluted ProcartaPlex Wash Buffer be stored?

The diluted buffer can be stored at 4 degrees C and should be used within 1 month of diluting.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can I get the catalog numbers for ProcartaPlex assay buffers and plates?

Here are the catalog numbers:

- EPX-11111-000: ProcartaPlex Universal Assay Buffer (1X) (10 mL) - It replaces all discontinued Procarta buffers (standard, assay, sample buffer); it is the same as the discontinued Procarta Cell Lysate Assay Buffer, 10 plate (Cat. No. PC0165), but only for one plate
- EPX-11110-000: ProcartaPlex Universal Assay Buffer (10X) (10 mL)
- EPXP-11112-000: ProcartaPlex Platinum Assay Buffer for Plasma Samples (1X, 10 mL)
- EPXP-11113-000: ProcartaPlex Platinum Assay Buffer for Serum Samples (1X, 10 mL)
- EPX-99999-000: ProcartaPlex Cell Lysis Buffer; It is the same as the discontinued Procarta Cell lysis Buffer (for cell lysate preparation) (Cat. No. PC6002)
- EPX-22222-000: ProcartaPlex Detection Antibody Diluent, Mouse (1X, 3 mL)
- EPX-33333-000: ProcartaPlex Detection Antibody Diluent, Human (1X, 3 mL)
- EPX-66666-000: ProcartaPlex Wash Buffer 10X (20 mL)
- EPX-66666-001: ProcartaPlex Wash Buffer 10X (200 mL)
- EPX-77777-000: ProcartaPlex Reading Buffer (1X, 40 mL)
- EPX-SAPE-000: ProcartaPlex Streptavidin-PE (1X, 5 mL) - 4.3 µg/mL (can range from 4.25-4.35 µg/mL)
- EPX-44444-000: ProcartaPlex 96 well Flat Bottom Plate, black (1 plate)
- EPX-88888-000: ProcartaPlex 96 well Filter Plate (1 plate)
- EPX-38488-000: ProcartaPlex 384 well Filter Plate (1 plate)

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.