ProcartaPlex™ Automation Setup Kit
ProcartaPlex™ Automation Setup Kit
ProcartaPlex™ Automation Setup Kit
Invitrogen™

ProcartaPlex™ Automation Setup Kit

The ProcartaPlex Automation Setup Kit enables setup, testing, and validation of instruments to automate crucial steps of the ProcartaPlex 384-well assay protocol.
Have Questions?
Catalog NumberQuantity
EPX-SETUP-3846 Tests
Catalog number EPX-SETUP-384
Price (INR)
63,959.00
Online offer
Ends: 31-Dec-2026
106,598.00
Save 42,639.00 (40%)
Each
Quantity:
6 Tests
Price (INR)
63,959.00
Online offer
Ends: 31-Dec-2026
106,598.00
Save 42,639.00 (40%)
Each

The ProcartaPlex Automation Setup Kit enables setup, testing, and validation of instruments to automate crucial steps of the ProcartaPlex 384-well assay protocol, such as liquid handling and plate washing, before running an actual assay with valuable sample. The user guide describes how to adjust and optimize an automated magnetic plate washer as needed during the ProcartaPlex 384-well assay workflow in order to ensure optimal performance and avoid bead loss.

Each kit provides six optimization tests for automated magnetic plate washing.

Specifications
For Use With (Equipment)FLEXMAP 3 D™ INTELLIFLEX™
Product LineProcartaPlex
Product TypeBasic and Setup Kits
Quantity6 Tests
Unit SizeEach
Contents & Storage

• Automation Setup Beads (1X)
• 1 x 200 mL Wash Buffer (10X)
• 1 Bottle Reading Buffer (1X)
• Adhesive film
• ProcartaPlex 384-well flat bottom plate

Store at 2°C to 8°C.