ProcartaPlex Mouse Basic Kit
Invitrogen™

ProcartaPlex Mouse Basic Kit

ProcartaPlex™ immunoassays are our ideal solution for assessing multiple protein biomarkers in a single sample.Our ProcartaPlex assays use LuminexRead more
Have Questions?
Catalog NumberQuantity
EPX010-20440-90196 Tests
Catalog number EPX010-20440-901
Price (INR)
-
Quantity:
96 Tests

ProcartaPlex™ immunoassays are our ideal solution for assessing multiple protein biomarkers in a single sample.

Our ProcartaPlex assays use Luminex ™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins.

ProcartaPlex™ assays are subject to the same stringent and robust development and manufacturing as are our extensively validated Platinum and Instant ELISA kits. Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma.Explore our broad portfolio of pre-configures panels and combinable single analytes.For combination of only Simplex Kits an additional Basic Kit is required. This kit includes all non-target specific reagents - assay diluent, wash buffers, SA-PE and plates.

This kit does not contain a Multiscreen Filter Plate.

Suitable Sample Types

Cell culture supernatant, serum, plasma

Sample Volume

Serum, Plasma: 25 μL; CCS: 50 μL

Reported Application

Multiplex Immunoassay

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay SensitivitySee Certificate of Analysis
For Use With (Equipment)Luminex™ Instruments
Product LineProcartaPlex
CombinabilityCombinable
Product TypeBasic and Setup Kits
Quantity96 Tests
SpeciesMouse
Unit SizeEach

Frequently asked questions (FAQs)

What is the size of the Luminex beads you currently use?

The beads used in our Luminex instrument-compatible ProcartaPlex and QuantiGene Plex assays are 6.5 micron superparamagnetic beads.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I am interested in performing Luminex assays using BioSource kits, and I have a Luminex xMAP system. Besides the kits and system, what other reagents and equipment will I need?

The following is a list of general lab supplies that are required for running BioSource immunoassays on the Luminex xMAP system:
1) Sonicating water bath
2) Orbital shaker
3) Vortexer
4) Repeating and/or multi-channel pipetter (not required, but recommended)
5) Calibrated adjustable precision pipettes, with disposable plastic tips
6) Glass/plastic tubes and racks for preparing reagents
7) Graduated cylinder and container for preparing wash solution
8) Aluminum foil
9) Deionized or distilled water.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Do the Luminex beads require special care in handling?

The Luminex beads should be protected from light because they are susceptible to photobleaching. We recommend protecting the beads by keeping containers covered with aluminum foil during all incubation steps, and exercising care during handling. The beads should not be frozen, subjected to excessive heat, or exposed to organic solvents.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Why would the Luminex acquisition software display "Sample Empty" messages during analysis?

(1) The user did not properly aliquot the diluted beads, such that no beads were actually added to the wells (make sure that the bead concentrates are sonicated and vortexed well, then check the pipet tip to ensure that air bubbles were not drawn up)
(2) The user missed loading diluted beads to some wells, which is likely since the small volume is clear and difficult to visualize in the clear plastic plate (we have now addressed this customer difficulty by coloring each of the Buffer Reagent Kit components)
(3) The user applied too much vacuum pressure at some point during the wash steps, or allowed the pressure to spike even once, such that the filter membrane tore in a few wells releasing the beads (make sure that the vacuum manifold pressure is kept below 5mm/in Hg, depending on their system -- a good rule of thumb is that it should take a full 3-second count to GENTLY empty the wells of 200uL)
(4) The user did not properly sonicate and vortex the beads prior to dilution, such that the percent of bead aggregation was high and the instrument was unable to find enough single beads to meet the events/bead value designated by the customer (make sure that the Bead Concentrate tube is put into the waterbath all the way to the cap, since the tube is hollow until the top third)
(5) The user lost beads by shaking the plate too aggressively or handling it improperly (make sure that the orbital shaker is set to a speed that allows for maximum vortex in the wells without spillage)
(6) The user exposed the beads to an excess of light during storage or running of the assay, such that some but not all of the beads were photobleached and therefore falling outside the acceptable range for each bead region (make sure that the plate is covered on the top/sides with foil throughout the assay, away from Windows and spotlights, and that the bead component of the kits is stored in the dark)*
(7) There was a clog in the sample needle, such that the instrument was unable to take up enough sample to meet the number of events requested per bead region (suggest that the user follow the manual instructions for dislodging a clog, which include several Back Flush steps and may require removal of the needle for sonication with probe alignment).

* Some of the older Antibody Bead Kits still have clear plastic tops instead of black ones. In cases where customers store kits in lit refrigerators, or keep them open on the lab bench, even a few hours of light exposure is enough to photobleach beads. It is important to note, in general, that higher number bead regions are more susceptible to photobleaching. In order to draw conclusions about the source of the difficulty, we would ask to see the data, specifically the Masterplex QT file, which would enable us to examine the pattern of "Sample Empty" occurrences in addition to the bead counts per well.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What are the Luminex beads made of?

The beads are made of polystyrene.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Citations & References (2)

Citations & References
Abstract
Diverse action of lipoteichoic acid and lipopolysaccharide on neuroinflammation, blood-brain barrier disruption, and anxiety in mice.
Authors:Mayerhofer R, Fröhlich EE, Reichmann F, Farzi A, Kogelnik N, Fröhlich E, Sattler W, Holzer P
Journal:Brain Behav Immun
PubMed ID:27751870
Microbial metabolites are known to affect immune system, brain, and behavior via activation of pattern recognition receptors such as Toll-like receptor 4 (TLR4). Unlike the effect of the TLR4 agonist lipopolysaccharide (LPS), the role of other TLR agonists in immune-brain communication is insufficiently understood. We therefore hypothesized that the TLR2 ... More
Intestinal Epithelial Cell Autophagy Is Required to Protect against TNF-Induced Apoptosis during Chronic Colitis in Mice.
Authors:Pott J, Kabat AM, Maloy KJ
Journal:Cell Host Microbe
PubMed ID:29358084
Genome-wide association studies have linked polymorphisms in the autophagy gene ATG16L1 with susceptibility to inflammatory bowel disease (IBD). However, the cell-type-specific effects of autophagy on the regulation of chronic intestinal inflammation have not been investigated. Here, we assessed the effect of myeloid-specific or intestinal epithelial cell (IEC)-specific deletion of Atg16l1 on ... More