mTOR Human ProcartaPlex™ Simplex Kit
mTOR Human ProcartaPlex™ Simplex Kit
Actual product may vary
Invitrogen™

mTOR Human ProcartaPlex™ Simplex Kit

The ProcartaPlex Human mTOR Simplex Kit measures the level of total mTOR in cell lysates. It is designed to be combinable with other Simplex kits so you can create your own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation.
Have Questions?
Catalog number EPX010-97005-901
Price (INR)
24,388.00
線上優惠
Ends: 31-Mar-2026
40,646.00
Save 16,258.00 (40%)
96 tests
Add to cart
Price (INR)
24,388.00
線上優惠
Ends: 31-Mar-2026
40,646.00
Save 16,258.00 (40%)
96 tests
Add to cart

The ProcartaPlex Human mTOR Simplex Kit measures the level of total mTOR in cell lysates. It is designed to be combinable with other Simplex kits so you can create your own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation. When combining multiple Simplex kits (i.e., when you are not using a pre-configured Multiplex Panel), only one buffer kit (sold separately) is needed for each assay plate regardless of plex size.

For preparation of cell lysates, we recommend use of either ProcartaPlex Cell Lysis Buffer (Cat. No. EPX-999999-901) or Cell Extraction Buffer (Cat. No. FNN0011) supplemented with 0.5 M EDTA and Halt Protease Inhibitor Cocktail (100X) (Cat. No. 78430). These buffers are not included with the panel. Depending on your cell system, other buffers may be suitable as well.

About ProcartaPlex Assays for the Luminex Platform
ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 µL of plasma or serum or 50 µL of cell culture supernatant and just four hours to obtain analyzed results.

More results per sample—measure multiple protein targets simultaneously in a single 25–50 µL sample
Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 µL sample from plasma, serum, cell culture supernatant, cell lysate, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g., Luminex 200, FLEXMAP 3D, xMAP INTELLIFLEX, xMAP INTELLIFLEX DR-SE, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

In addition, the xMAP INTELLIFLEX DR-SE System offers dual-reporter capability, allowing the measurement of a second parameter of the analyte, and features side eject functionalities for added flexibility and automation. Together with ProcartaPlex signaling assays, the xMAP INTELLIFLEX DR-SE instrument enables the quantitation and multiplexing of phosphorylated and total proteins in the same well, on the same Luminex bead, and in the same assay run.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

The ProcartaPlex Human mTOR Simplex Kit measures the level of total mTOR in cell lysates. It is designed to be combinable with other Simplex kits so you can create your own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation. When combining multiple Simplex kits (i.e., when you are not using a pre-configured Multiplex Panel), only one buffer kit (sold separately) is needed for each assay plate regardless of plex size.

For preparation of cell lysates, we recommend use of either ProcartaPlex Cell Lysis Buffer (Cat. No. EPX-999999-901) or Cell Extraction Buffer (Cat. No. FNN0011) supplemented with 0.5 M EDTA and Halt Protease Inhibitor Cocktail (100X) (Cat. No. 78430). These buffers are not included with the panel. Depending on your cell system, other buffers may be suitable as well.

About ProcartaPlex Assays for the Luminex Platform
ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 µL of plasma or serum or 50 µL of cell culture supernatant and just four hours to obtain analyzed results.

More results per sample—measure multiple protein targets simultaneously in a single 25–50 µL sample
Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 µL sample from plasma, serum, cell culture supernatant, cell lysate, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g., Luminex 200, FLEXMAP 3D, xMAP INTELLIFLEX, xMAP INTELLIFLEX DR-SE, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

In addition, the xMAP INTELLIFLEX DR-SE System offers dual-reporter capability, allowing the measurement of a second parameter of the analyte, and features side eject functionalities for added flexibility and automation. Together with ProcartaPlex signaling assays, the xMAP INTELLIFLEX DR-SE instrument enables the quantitation and multiplexing of phosphorylated and total proteins in the same well, on the same Luminex bead, and in the same assay run.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

For Research Use Only.
Specifications
Product TypeSimplex Kit
Quantity96 Tests
SpeciesHuman
CombinabilityCombinable
Bead TypeMagnetic
Assay SensitivityAs determined for Lot 1: 0.74 U/mL
Assay RangeAs determined for Lot 1: 2.44 to 10,000 U/mL
FormatSimplex Kit
For Use With (Equipment)Luminex™ Instruments
GenemTOR
Gene AliasMTOR, FRAP, FRAP1, FRAP2, RAFT1, RAPT1, SKS, mechanistic target of rapamycin, mechanistic target of rapamycin kinase
Gene ID (Entrez)2475
Gene SymbolmTOR
Interassay CVAs determined for Lot 1: 7.63%
Intraassay CVAs determined for Lot 1: 8.71%
Product LineProcartaPlex™
Sample TypeCell lysates
Sample VolumeCell lysates: 50 μL
UniProt IDP42345
Shipping ConditionWet Ice
Research AreaSignaling
Unit Size96 tests