MEK1[pS218/pS222] Human ProcartaPlex™ Simplex Kit
Invitrogen™

MEK1[pS218/pS222] Human ProcartaPlex™ Simplex Kit

Invitrogen ProcartaPlex Human MEK1[pS218/pS222] Simplex Kit enables precise analysis of the Protein of Interest.
Have Questions?
Catalog NumberQuantity
EPX010-97114-PHO96 Tests
Catalog number EPX010-97114-PHO
Price (INR)
44,050.00
Each
Quantity:
96 Tests
Price (INR)
44,050.00
Each
The ProcartaPlex Human MEK1[pS218/pS222] Simplex Kit measures the level of phosphorylated MEK1[pS218/pS222] in cell lysates. It is designed to be combinable with other Simplex kits so you can create your own multiplex panel that utilizes Luminex xMAP technology for protein quantitation. When combining multiple Simplex kits (i.e., when you are not using a pre-configured Multiplex Panel), only one buffer kit (sold separately) is needed for each assay plate regardless of plex size.

For preparation of cell lysates, we recommend the use of either ProcartaPlex Cell Lysis Buffer (Cat. No. EPX-99999-000) or Cell Extraction Buffer (Cat. No. FNN0011) supplemented with 0.5 M EDTA and Halt Protease and Phosphatase Inhibitor Single to Use Cocktail 100X (Cat. No. 78440 or 78442). These buffers are not included in the kit. Depending on your cell system, other buffers can be suitable as well.

The ProcartaPlex Human MEK1[pS218/pS222] Simplex Kit is part of the ProcartaPlex Human MAPK Pathway Panel (phospho) 6plex (Cat. No. EPX060-97117-PHO), which complements the ProcartaPlex Human MAPK Pathway Panel (total) 6plex (Cat. No. EPX060-97017-901).

For Research Use Only.
Specifications
Assay RangeSee Certificate of Analysis
Assay SensitivitySee Certificate of Analysis
Bead RegionMEK1[pS218/pS222] [63]
Bead TypeMagnetic
For Use With (Equipment)Luminex™ Instruments
GeneMAP2K1
Gene Alias5604
Gene ID (Entrez)MAP2K1
High-throughput CompatibilityYes
Interassay CVSee Certificate of Analysis
Intraassay CVSee Certificate of Analysis
Product LineProcartaPlex
Sample TypeCell lysates
Sample VolumeCell lysates (prediluted): 25 μL
Shipping ConditionWet Ice
SynonymMitogen-activated protein kinase
UniProt IDQ02750
CombinabilityCombinable
Product TypeSimplex Kit
Quantity96 Tests
Research AreaSignaling
SpeciesHuman
Unit SizeEach
Contents & Storage
1 vial Capture Beads (50x)
1 vial biotinylated Detection Antibody (50x)
2 vials Standard Mix (lyophilized)

Store as indicated on the Certificate of Analysis.