STAT5A[pY694] Human ProcartaPlex™ Simplex Kit
Invitrogen™

STAT5A[pY694] Human ProcartaPlex™ Simplex Kit

ProcartaPlex Simplex kits enable precise analysis of the protein of interest.
Have Questions?
Catalog NumberQuantity
EPX010-97125-PHO96 Tests
Catalog number EPX010-97125-PHO
Price (INR)
29,611.00
Offre exceptionnelle en ligne
Ends: 31-Mar-2026
49,351.00
Save 19,740.00 (40%)
Each
Quantity:
96 Tests
Price (INR)
29,611.00
Offre exceptionnelle en ligne
Ends: 31-Mar-2026
49,351.00
Save 19,740.00 (40%)
Each

The ProcartaPlex Human STAT5A[pY694] Simplex Kit measures the level of total STAT5A[pY694] in cell lysates. It is designed to be combinable with other Simplex kits so you can create your own multiplex panel that utilizes Luminex xMAP technology for protein quantitation. When combining multiple Simplex kits (i.e., when you are not using a pre-configured Multiplex Panel), only one buffer kit (sold separately) is needed for each assay plate regardless of plex size.

For preparation of cell lysates, we recommend the use of either ProcartaPlex Cell Lysis Buffer (Cat. No. EPX-99999-000) or Cell Extraction Buffer (Cat. No. FNN0011) supplemented with 0.5 M EDTA and Halt Protease and Phosphatase Inhibitor Single–Use Cocktail 100X (Cat. No. 78440 or 78442). These buffers are not included with the kit. Depending on your cell system, other buffers can be suitable as well.

The ProcartaPlex Human STAT5A[pY694] Simplex Kit is part of the ProcartaPlex Human Multi Pathway Panel (phospho), 12plex (Cat. No. EPX120-97130-PHO), which complements the ProcartaPlex Human Multi Pathway Panel (total), 12plex (Cat. No. EPX120-97030-901).

In addition to the STAT5A[pY694] Simplex Kit we also offer a STAT5A (total) Simplex Kit as well as kits to measure other targets in the JAK-STAT signaling pathway, such as STAT1, STAT1[pY701], STAT3, STAT3[pY705], STAT6, STAT6[pY641], JAK2, JAK2[pY1007/1008], FYN, FYN[pY530], SRC, SRC[pY416].

About ProcartaPlex Assays for the Luminex Platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

  • More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
  • Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g. Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine). Visit thermofisher.com/procartaplex for more information, including a comprehensive list of individual protein targets.

For Research Use Only.
Specifications
Assay RangeSee Certificate of Analysis
Assay SensitivitySee Certificate of Analysis
Bead RegionSTAT5A[pY694] [51]
Bead TypeMagnetic
For Use With (Equipment)Luminex™ Instruments
FormatSimplex Kit
GeneSTAT5A
Gene AliasSignal Transducer And Activator Of Transcription 5A
Gene ID (Entrez)6776
Gene SymbolSTAT5A
Interassay CVSee Certificate of Analysis
Intraassay CVSee Certificate of Analysis
Multiplex Size Plex No.Simplex
Product LineProcartaPlex
Sample TypeCell lysates
Sample VolumeCell lysates (prediluted): 25 μL
Shipping ConditionWet Ice
SynonymSignal Transducer And Activator Of Transcription 5A, Epididymis Secretory Sperm Binding Protein
UniProt IDP42229
CombinabilityCombinable
Product TypeSimplex Kit
Quantity96 Tests
Research AreaSignaling
SpeciesHuman
Unit SizeEach
Contents & Storage
See Certificate of Analysis