IL-2 Rat ProcartaPlex™ Simplex Kit
Invitrogen™

IL-2 Rat ProcartaPlex™ Simplex Kit

The Rat IL-2 Simplex ProcartaPlex Kit measures IL-2 protein and is designed to be combinable with other Simplex kits so that you can create your own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation.
Have Questions?
Catalog NumberQuantity
EPX01A-36052-90196 Tests
Catalog number EPX01A-36052-901
Price (INR)
-
Quantity:
96 Tests

The Rat IL-2 Simplex ProcartaPlex Kit measures IL-2 protein and is designed to be combinable with other Simplex kits so that you can create your own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation. When combining multiple Simplex kits (i.e., when you are not using a pre-configured Multiplex Panel), only one buffer kit (sold separately) is needed for each assay plate regardless of plex size.

About ProcartaPlex Assays for the Luminex Platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

  • More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
  • Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g. Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine). Visit thermofisher.com/procartaplex for more information, including a comprehensive list of individual protein targets.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay RangeAs determined for Lot 1: 4.9 - 20,000 pg/mL
Assay SensitivityAs determined for Lot 1: 1.9 pg/mL
Bead TypeIL-2 [20]
For Use With (Equipment)Luminex™ Instruments
FormatSimplex Kit
GeneInterleukin 2
Gene ID (Entrez)116562
Gene SymbolIL2
Product LineProcartaPlex
ProteinInterleukin-2
Protein SubtypeIL-2, T-cell growth factor, TCGF
Sample TypeSerum, Plasma, Cell Culture Supernatants
Sample VolumeSerum, Plasma: 25 μL, CCS: 50 μL
Shipping ConditionWet Ice
UniProt IDP17108
CombinabilityCombinable
Product TypeSimplex Kit
Quantity96 Tests
Research AreaImmunology, Oncology, Neurobiology, Toxicology
SpeciesRat
Unit SizeEach
Contents & Storage

• 1 vial Capture Beads (50X)
• 1 vial biotinylated Detection Antibody (50X)
• 2 vials Rat Standard Mix A (lyophilized)

Store at 2°C to 8°C.

Frequently asked questions (FAQs)

What is the size of the Luminex beads you currently use?

The beads used in our Luminex instrument-compatible ProcartaPlex and QuantiGene Plex assays are 6.5 micron superparamagnetic beads.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I am interested in performing Luminex assays using BioSource kits, and I have a Luminex xMAP system. Besides the kits and system, what other reagents and equipment will I need?

The following is a list of general lab supplies that are required for running BioSource immunoassays on the Luminex xMAP system:
1) Sonicating water bath
2) Orbital shaker
3) Vortexer
4) Repeating and/or multi-channel pipetter (not required, but recommended)
5) Calibrated adjustable precision pipettes, with disposable plastic tips
6) Glass/plastic tubes and racks for preparing reagents
7) Graduated cylinder and container for preparing wash solution
8) Aluminum foil
9) Deionized or distilled water.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Do the Luminex beads require special care in handling?

The Luminex beads should be protected from light because they are susceptible to photobleaching. We recommend protecting the beads by keeping containers covered with aluminum foil during all incubation steps, and exercising care during handling. The beads should not be frozen, subjected to excessive heat, or exposed to organic solvents.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Why would the Luminex acquisition software display "Sample Empty" messages during analysis?

(1) The user did not properly aliquot the diluted beads, such that no beads were actually added to the wells (make sure that the bead concentrates are sonicated and vortexed well, then check the pipet tip to ensure that air bubbles were not drawn up)
(2) The user missed loading diluted beads to some wells, which is likely since the small volume is clear and difficult to visualize in the clear plastic plate (we have now addressed this customer difficulty by coloring each of the Buffer Reagent Kit components)
(3) The user applied too much vacuum pressure at some point during the wash steps, or allowed the pressure to spike even once, such that the filter membrane tore in a few wells releasing the beads (make sure that the vacuum manifold pressure is kept below 5mm/in Hg, depending on their system -- a good rule of thumb is that it should take a full 3-second count to GENTLY empty the wells of 200uL)
(4) The user did not properly sonicate and vortex the beads prior to dilution, such that the percent of bead aggregation was high and the instrument was unable to find enough single beads to meet the events/bead value designated by the customer (make sure that the Bead Concentrate tube is put into the waterbath all the way to the cap, since the tube is hollow until the top third)
(5) The user lost beads by shaking the plate too aggressively or handling it improperly (make sure that the orbital shaker is set to a speed that allows for maximum vortex in the wells without spillage)
(6) The user exposed the beads to an excess of light during storage or running of the assay, such that some but not all of the beads were photobleached and therefore falling outside the acceptable range for each bead region (make sure that the plate is covered on the top/sides with foil throughout the assay, away from Windows and spotlights, and that the bead component of the kits is stored in the dark)*
(7) There was a clog in the sample needle, such that the instrument was unable to take up enough sample to meet the number of events requested per bead region (suggest that the user follow the manual instructions for dislodging a clog, which include several Back Flush steps and may require removal of the needle for sonication with probe alignment).

* Some of the older Antibody Bead Kits still have clear plastic tops instead of black ones. In cases where customers store kits in lit refrigerators, or keep them open on the lab bench, even a few hours of light exposure is enough to photobleach beads. It is important to note, in general, that higher number bead regions are more susceptible to photobleaching. In order to draw conclusions about the source of the difficulty, we would ask to see the data, specifically the Masterplex QT file, which would enable us to examine the pattern of "Sample Empty" occurrences in addition to the bead counts per well.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What are the Luminex beads made of?

The beads are made of polystyrene.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Citations & References (1)

Citations & References
Abstract
Enhancement of immune maturation in suckling rats by leptin and adiponectin supplementation.
Authors:Grases-Pintó B, Abril-Gil M, Castell M, Pérez-Cano FJ, Franch À
Journal:Sci Rep
PubMed ID:30742004
'Leptin and adiponectin, adipokines present in breast milk, have shown immunomodulatory properties. The current study aimed to ascertain whether a nutritional supplementation with leptin or adiponectin in neonatal rats was able to influence the maturation of the systemic immune response in early life. To achieve this, suckling Wistar rats were ... More