ProcartaPlex™ Human Brain Injury Panel, 5plex
Invitrogen™

ProcartaPlex™ Human Brain Injury Panel, 5plex

The ProcartaPlex Human Brain Injury Panel 5plex helps enable neurological research by analyzing the protein targets GFAP, NF-H, S100B, NSE, and UCHL1 in a single well using Luminex xMAP technology.
Have Questions?
Catalog NumberQuantity
EPX050-15838-90196 Tests
Catalog number EPX050-15838-901
Price (INR)
128,170.00
96 tests
Quantity:
96 Tests
Price (INR)
128,170.00
96 tests

The ProcartaPlex Human Brain Injury Panel 5plex helps enable neurological research by analyzing the protein targets GFAP, NF-H, S100B, NSE, and UCHL1 in a single well using Luminex xMAP technology. Simultaneous detection of these targets, released due to astroglial injury (GFAP, S100B), axonal injury (NF-H), and neuronal cell body injury (UCH-L1), can provide a better understanding of the progression of neurological disorders.

The panel is provided in a ready-to-use format with individual vials of capture and detection reagents formulated at 1X concentration, requiring less pipetting and experimental setup. It is not combinable with simplexes or other panels. It complements the Human Neuroinflammation Panel 6plex (Cat. No. EPX060-15833-901) and Human Neurodegeneration Panel 1 9plex (Cat. No. EPX090-15836-901), providing 15 additional protein targets in the same research area. ProcartaPlex preconfigured panels are extensively tested for analyte combinability, interference, and cross-reactivity to provide the highest level of validation and precision.

Target list [bead region]: GFAP [22], NF-H [33], NSE [12], S100B [18], UCHL1 [19]

About ProcartaPlex assays for the Luminex platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

  • Reproducible, reliable results—validated as a panel to the highest industry standard, including protein target combinability and cross-reactivity testing
  • More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
  • Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample—from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g., Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay RangeSee Certificate of Analysis
Assay SensitivitySee Certificate of Analysis
Bead RegionNSE [12], GFAP [22], pNF-H [33], UCHL-1 [19], S100b [18]
Bead TypeMagnetic
For Use With (Equipment)Luminex™ Instruments
FormatMultiplex Kit
Interassay CVLess than 15%
Intraassay CVLess than 15%
Product LineProcartaPlex
Sample TypeSerum, Plasma, Cerebrospinal Fluid
Sample VolumeSerum, Plasma, CSF: 25 μL
Shipping ConditionWet Ice
CombinabilityNon-Combinable
Product TypeMultiplex Panel
Quantity96 Tests
Research AreaNeurobiology
SpeciesHuman
Unit Size96 tests
Contents & Storage

• 2 vials Human Standard Mix 4 (lyophilized)
• 1 vial Capture Bead Mix (1X)
• 1 vial Biotinylated Detection Antibody Mix (1X)
• 1 bottle Reading Buffer (1X)
• 1 bottle Streptavidin-PE (1X)
• 1 bottle Universal Assay Buffer (1X)
• 1 bottle Wash Buffer (10X)
• 8-tube strip
• Adhesive film
• Flat-bottom 96-well plate, black

Store at 2°C to 8°C.