ProcartaPlex Human Multi Pathway Panel (phospho), 12plex
Invitrogen™
ProcartaPlex Human Multi Pathway Panel (phospho), 12plex
ProcartaPlex panels enable precise analysis of the proteins of interest.
Have Questions?
Catalog NumberQuantity
EPX120-97130-PHO96 Tests
Catalog number EPX120-97130-PHO
Price (INR)
214,099.00
線上優惠
Ends: 31-Mar-2026
356,831.00
Save 142,732.00 (40%)
Each
Quantity:
96 Tests
Price (INR)
214,099.00
線上優惠
Ends: 31-Mar-2026
356,831.00
Save 142,732.00 (40%)
Each
The ProcartaPlex Human Multi Pathway Panel (phospho), 12plex, enables simultaneous quantitation of the total levels of twelve phosphorylated key proteins iin various cell signaling pathways. It complements the ProcartaPlex Human Multi Pathway Panel (total), 12plex (Cat. No. EPX120-97130-901).
The panel is validated for use with cell lysates. For preparation of cell lysates, we recommend the use of either ProcartaPlex Cell Lysis Buffer (Cat. No. EPX-99999-000) or Cell Extraction Buffer (Cat. No. FNN0011) supplemented with 0.5 M EDTA and Halt Protease and Phosphatase Inhibitor Single–Use Cocktail 100X (Cat. No. 78440 or 78442). These buffers are not included in the panel. Depending on your cell system, other buffers can be suitable as well.
This kit is compatible with all Luminex instruments. If you use an xMAP INTELLIFLEX DR-SE instrument, we recommend our ProcartaPlex Human Multi Pathway Dual Reporter Panel, 2x12plex (Cat. No. EPX120-97230-DR), which enables the simultaneous quantitation of total and phosphorylated proteins on the same bead in a single well.
The panel is provided in a ready-to-use format with individual vials of capture and detection reagents formulated at 1X concentration, requiring less pipetting and experimental setup.
ProcartaPlex preconfigured panels are extensively tested for analyte combinability, interference, and cross-reactivity to provide the highest level of validation and precision. All ProcartaPlex panels are supplied with the necessary reagents to perform the assay.
Targets [bead region]: Akt[pS473] [47], CREB [PS133] [43], mTOR[Ser2448] [26], p70S6K[pTpS421/424] [27], NFkBp65[Ser536] [15], STAT5A[pY694] [51], STAT6[pY641] [75], JAK2[pY1007/1008] [55], JNK1/3[pT183/pT221] [13], FAK[pY397] [30], c-Myc[pS62] [53], ERK1[pT202/pY204] [62]
About ProcartaPlex Assays for the Luminex Platform
ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.
  • More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
  • Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation
ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample from plasma, serum, cell culture supernatants, and other bodily fluids.
The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g. Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.
The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.
ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine). Visit thermofisher.com/procartaplex for more information, including a comprehensive list of individual protein targets.
For Research Use Only.
Specifications
Bead TypeMagnetic
For Use With (Equipment)Luminex™ Instruments
FormatMultiplex Kit
Multiplex Size Plex No.Panel
Product LineProcartaPlex
Sample TypeCell lysates
Sample VolumeCell lysates (prediluted): 25 μL
Shipping ConditionWet Ice
CombinabilityNon-Combinable
Product TypeMultiplex Panel
Quantity96 Tests
Research AreaSignaling
SpeciesHuman
Unit SizeEach
Contents & Storage
See Certificate of Analysis