ProcartaPlex Human Multi Pathway Dual Reporter Panel, 2x12plex
Invitrogen™
ProcartaPlex Human Multi Pathway Dual Reporter Panel, 2x12plex
ProcartaPlex panels enable precise analysis of the proteins of interest.
Have Questions?
Catalog NumberQuantity
EPX120-97230-DR96 Tests
Catalog number EPX120-97230-DR
Price (INR)
397,519.00
Online offer
Ends: 31-Mar-2026
662,531.00
Save 265,012.00 (40%)
Each
Quantity:
96 Tests
Price (INR)
397,519.00
Online offer
Ends: 31-Mar-2026
662,531.00
Save 265,012.00 (40%)
Each
The ProcartaPlex Human Multi Pathway Dual Reporter Panel, 2x12plex, enables simultaneous quantitation of 24 parameters (12 total and 12 phosphorylated proteins) on the same bead in a single well. This is realized by leveraging two distinct detector antibodies with different dye-conjugates in the assay and the dual reporter capabilities and additional violet laser of the xMAP INTELLIFLEX DR-SE instrument.
The ProcartaPlex Human Multi Pathway Dual Reporter Panel, 2x12plex, is validated for use with cell lysates. For preparation of cell lysates, we recommend the use of either ProcartaPlex Cell Lysis Buffer (Cat. No. EPX-99999-000) or Cell Extraction Buffer (Cat. No. FNN0011) supplemented with 0.5 M EDTA and Halt Protease and Phosphatase Inhibitor Single–Use Cocktail 100X (Cat. No. 78440 or 78442). These buffers are not included in the kit. Depending on your cell system, other buffers can be suitable as well.
The panel is provided in a ready-to-use format with individual vials of capture and detection reagents formulated at 1X concentration, requiring less pipetting and experimental setup.
ProcartaPlex preconfigured panels are extensively tested for analyte combinability, interference, and cross-reactivity to provide the highest level of validation and precision. All ProcartaPlex panels are supplied with the necessary reagents to perform the assay.
This kit is only compatible with the xMAP INTELLIFLEX DR-SE instrument. For all other Luminex instruments, we offer ProcartaPlex Human Multi Pathway Panel (total), 12plex (Cat. No. EPX120-97030-901) and ProcartaPlex Human Multi Pathway Panel (phospho), 12plex (Cat. No. EPX120-97130-PHO). Other options include the purchase of Simplex kits or the use of our Panel Configurator to create a custom Mix&Match panel.
Targets [bead region]: Akt + Akt[pS473] [47], CREB + CREB[PS133] [43], mTOR + mTOR[Ser2448] [26], p70S6K + p70S6K[pTpS421/424] [27], NFkBp65 + NFkBp65[Ser536] [15], STAT5A + STAT5A[pY694] [51], STAT6 + STAT6[pY641] [75], JAK2 + JAK2[pY1007/1008] [55], JNK1/3 + JNK1/3[pT183/pT221] [13], FAK + FAK[pY397] [30], c-Myc + c-Myc[pS62] [53], ERK1 + ERK1[pT202/pY204] [62]
About ProcartaPlex Assays for the Luminex Platform
ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.
  • More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
  • Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation
ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample from plasma, serum, cell culture supernatants, and other bodily fluids.
The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g. Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.
The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.
ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine). Visit thermofisher.com/procartaplex for more information, including a comprehensive list of individual protein targets.
For Research Use Only.
Specifications
Bead TypeMagnetic
For Use With (Equipment)Luminex™ Instruments
FormatMulitplex Kit
Multiplex Size Plex No.Panel
Product LineProcartaPlex
Sample TypeCell lysates
Sample VolumeCell lysates (prediluted): 25 μL
Shipping ConditionWet Ice
CombinabilityNon-Combinable
Product TypeMultiplex Panel
Quantity96 Tests
Research AreaSignaling
SpeciesHuman
Unit SizeEach
Contents & Storage
See Certificate of Analysis