ProcartaPlex™ Human Neuroscience Panel, 21plex
Invitrogen™

ProcartaPlex™ Human Neuroscience Panel, 21plex

The ProcartaPlex Human Neuroscience Panel, 21plex, is the biggest ProcartaPlex panel for neurological research and enables the quantification of 21 protein targets in a single well using Luminex xMAP technology.
Have Questions?
Catalog NumberQuantity
EPX210-15839-90196 Tests
Catalog number EPX210-15839-901
Price (INR)
533,071.00
Each
Quantity:
96 Tests
Price (INR)
533,071.00
Each

The ProcartaPlex Human Neuroscience Panel, 21plex, is the biggest ProcartaPlex panel for neurological research and enables the quantification of 21 protein targets in a single well using Luminex xMAP technology. The panel is provided in a ready-to-use format with individual vials of capture and detection reagents formulated at 1X concentration, requiring less pipetting and experimental setup. It is not combinable with simplexes or other panels.

ProcartaPlex preconfigured panels are extensively tested for analyte combinability, interference, and cross-reactivity to provide the highest level of validation and precision. ProcartaPlex panels are supplied with the necessary reagents to perform the assay.

Target list [bead region]: amyloid beta 1-42 [42], alpha-synuclein (total) [53], BDNF [57], CNTF [36], FGF-21 [76], GDNF [44], GFAP [22], kallikrein-6 (KLK6) [14], NCAM-1 [48], neurogranin (NRGN) [75], NF-H [33], NF-L [78], NGF beta [55], NSE [12] S100B [18], TAU (total) [46], TAU [p181] [61], TDP-43 [74], TREM-2 [13] UCHL-1 [19], and YKL-40 (CHI3L1) [34]

About ProcartaPlex assays for the Luminex platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum or 50 μL of cell culture supernatant and just four hours to obtain analyzed results.

Features include:

  • Reproducible, reliable results—validated as a panel to the highest industry standard, including protein target combinability and cross-reactivity testing
  • More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
  • Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g., Luminex 200, FLEXMAP 3D, xMAP INTELLIFLEX, xMAP INTELLIFLEX DR-SE, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine).

For Research Use Only.
Specifications
Assay RangeSee Certificate of Analysis
Assay SensitivitySee Certificate of Analysis
For Use With (Equipment)Luminex™ Instruments
Interassay CVLess than 15%
Intraassay CVLess than 15%
Multiplex Size Plex No.21 plex
Product LineProcartaPlex
Sample TypeSerum, Plasma, Cerebrospinal Fluid
Sample VolumeSerum, Plasma, CSF: 25 μL
Shipping ConditionWet Ice
CombinabilityNon-Combinable
Product TypeMultiplex Panel
Quantity96 Tests
Research AreaNeurobiology
SpeciesHuman
Unit SizeEach