ProcartaPlex™ Human Immune Monitoring Panel for MAGPIX, 65plex
Invitrogen™

ProcartaPlex™ Human Immune Monitoring Panel for MAGPIX, 65plex

The ProcartaPlex Human Immune Monitoring Panel for MAGPIX 65plex is a preconfigured multiplex immunoassay kit that measures 65 cytokines, chemokines, and growth factors using Luminex xMAP technology.
Have Questions?
Catalog NumberQuantity
EPX650-16500-90196 Tests
Catalog number EPX650-16500-901
Price (INR)
1,000,839.00
Each
Quantity:
96 Tests
Price (INR)
1,000,839.00
Each

The ProcartaPlex Human Immune Monitoring Panel for MAGPIX 65plex is a preconfigured multiplex immunoassay kit that measures 65 cytokines, chemokines, and growth factors using Luminex xMAP technology. Analyze these 65 targets simultaneously for efficient immune response profiling, biomarker discovery, and validation. This kit consists of two separate kits: a 43-plex and a 22-plex kit that have been specifically designed to be used on the MAGPIX instrument that is capable of detecting up to 50 targets simultaneously. Luminex 100 and FLEXMAP 3D users should use the Human Immune Monitoring Panel 65plex (Cat. No. EPX650-10065-901) that contains all 65 targets in a single panel. The panels (43-plex and 22-plex) are provided in a ready-to-use format with individual vials of capture and detection reagents formulated at 1X concentration, requiring less pipetting and experimental setup.

ProcartaPlex preconfigured panels are extensively tested for analyte combinability, interference, and cross-reactivity to provide the highest level of validation and precision. All ProcartaPlex panels are supplied with the necessary reagents to perform the assay.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine).

Target list [bead region]:

  • 22-plex: APRIL [52], BAFF [65], BLC (CXCL13) [15], CD30 [34], ENA-78 (CXCL5) [55], Eotaxin-2 (CCL24) [37], Eotaxin-3 (CCL26) [77], FGF-2 [75], Fractalkine (CX3CL1) [39], IL-16 [47], IL-2R (CD25) [22], IL-20 [26], I-TAC (CXCL11) [46], MCP-2 (CCL8) [18], MCP-3 (CCL7) [66], MDC (CCL22) [30], MIF [67], MIG (CXCL9) [54], TNF-RII [45], TRAIL (CD253) [29], TSLP [61], TWEAK [35]
  • 43-plex: CD40L (CD154) [74], Eotaxin (CCL11) [33], Gro-alpha (CXCL1) [61], G-CSF (CSF-3) [42], GM-CSF [44], HGF [46], IFN alpha [48], IP-10 (CXCL10) [22], IFN gamma [43], IL-1 alpha [62], IL-1 beta [18], IL-2 [19], IL-3 [73], IL-4 [20], IL-5 [21], IL-6 [25], IL-7 [26], IL-8 (CXCL8) [27], IL-9 [52], IL-10 [28], IL-12p70 [34], IL-13 [35], IL-15 [65], IL-17A (CTLA-8) [36], IL-18 [66], IL-21 [72], IL-22 [76], IL-23 [63], IL-27 [14], IL-31 [37], LIF [15], M-CSF [67], MCP-1 (CCL2) [51], MIP-1 alpha (CCL3) [12], MIP-1 beta (CCL4) [47], MIP-3 alpha (CCL20) [56], MMP-1 [64], NGF beta [55], SDF-1 alpha (CXCL12) [13], SCF [39], TNF alpha [45], TNF beta [54], VEGF-A [78]

About ProcartaPlex assays for the Luminex platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

  • Reproducible, reliable results—validated as a panel to the highest industry standard, including protein target combinability and cross-reactivity testing
  • More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
  • Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25 to 50 μL sample—from plasma, serum, cell culture supernatants, and other bodily fluids.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay RangeSee Certificate of Analysis
Assay SensitivitySee Certificate of Analysis
Detection MethodFluorescence
For Use With (Equipment)Luminex™ Instruments
FormatMultiplex Kit
Product LineProcartaPlex
Sample TypeSerum, Plasma, Cell Culture Supernatants
Sample VolumeSerum, Plasma: 25 μL, CCS: 50 μL
Shipping ConditionWet Ice
CombinabilityNon-Combinable
Product TypeMultiplex Panel
Quantity96 Tests
Research AreaImmunology
SpeciesHuman
Unit SizeEach
Contents & Storage

• 4 vials Human Standard Mix M (lyophilized)
• 1 vial Capture Bead Mix M1 (1X)
• 1 vial Capture Bead Mix M2 (1X)
• 1 vial Biotinylated Detection Antibody Mix M1 (1X)
• 1 vial Biotinylated Detection Antibody Mix M2 (1X)
• 4 vials Human Control-High
• 4 vials Human Control-Low
• 1 Bottle Reading Buffer (1X)
• 1 Bottle Wash Buffer (10X)
• 1 Bottle Streptavidin-PE (1X)
• 1 Bottle Universal Assay Buffer (1X)
• 8-tube strip
• Adhesive film
• 2 Flat-bottom 96-well plate, black

Store at 2°C to 8°C

Frequently asked questions (FAQs)

What is the size of the Luminex beads you currently use?

The beads used in our Luminex instrument-compatible ProcartaPlex and QuantiGene Plex assays are 6.5 micron superparamagnetic beads.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I am interested in performing Luminex assays using BioSource kits, and I have a Luminex xMAP system. Besides the kits and system, what other reagents and equipment will I need?

The following is a list of general lab supplies that are required for running BioSource immunoassays on the Luminex xMAP system:
1) Sonicating water bath
2) Orbital shaker
3) Vortexer
4) Repeating and/or multi-channel pipetter (not required, but recommended)
5) Calibrated adjustable precision pipettes, with disposable plastic tips
6) Glass/plastic tubes and racks for preparing reagents
7) Graduated cylinder and container for preparing wash solution
8) Aluminum foil
9) Deionized or distilled water.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Do the Luminex beads require special care in handling?

The Luminex beads should be protected from light because they are susceptible to photobleaching. We recommend protecting the beads by keeping containers covered with aluminum foil during all incubation steps, and exercising care during handling. The beads should not be frozen, subjected to excessive heat, or exposed to organic solvents.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Why would the Luminex acquisition software display "Sample Empty" messages during analysis?

(1) The user did not properly aliquot the diluted beads, such that no beads were actually added to the wells (make sure that the bead concentrates are sonicated and vortexed well, then check the pipet tip to ensure that air bubbles were not drawn up)
(2) The user missed loading diluted beads to some wells, which is likely since the small volume is clear and difficult to visualize in the clear plastic plate (we have now addressed this customer difficulty by coloring each of the Buffer Reagent Kit components)
(3) The user applied too much vacuum pressure at some point during the wash steps, or allowed the pressure to spike even once, such that the filter membrane tore in a few wells releasing the beads (make sure that the vacuum manifold pressure is kept below 5mm/in Hg, depending on their system -- a good rule of thumb is that it should take a full 3-second count to GENTLY empty the wells of 200uL)
(4) The user did not properly sonicate and vortex the beads prior to dilution, such that the percent of bead aggregation was high and the instrument was unable to find enough single beads to meet the events/bead value designated by the customer (make sure that the Bead Concentrate tube is put into the waterbath all the way to the cap, since the tube is hollow until the top third)
(5) The user lost beads by shaking the plate too aggressively or handling it improperly (make sure that the orbital shaker is set to a speed that allows for maximum vortex in the wells without spillage)
(6) The user exposed the beads to an excess of light during storage or running of the assay, such that some but not all of the beads were photobleached and therefore falling outside the acceptable range for each bead region (make sure that the plate is covered on the top/sides with foil throughout the assay, away from Windows and spotlights, and that the bead component of the kits is stored in the dark)*
(7) There was a clog in the sample needle, such that the instrument was unable to take up enough sample to meet the number of events requested per bead region (suggest that the user follow the manual instructions for dislodging a clog, which include several Back Flush steps and may require removal of the needle for sonication with probe alignment).

* Some of the older Antibody Bead Kits still have clear plastic tops instead of black ones. In cases where customers store kits in lit refrigerators, or keep them open on the lab bench, even a few hours of light exposure is enough to photobleach beads. It is important to note, in general, that higher number bead regions are more susceptible to photobleaching. In order to draw conclusions about the source of the difficulty, we would ask to see the data, specifically the Masterplex QT file, which would enable us to examine the pattern of "Sample Empty" occurrences in addition to the bead counts per well.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What are the Luminex beads made of?

The beads are made of polystyrene.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Citations & References (1)

Citations & References
Abstract
Multiplexing protein and gene level measurements on a single Luminex platform.
Authors:Cook DB, McLucas BC, Montoya LA, Brotski CM, Das S, Miholits M, Sebata TH
Journal:Methods
PubMed ID:30742996
'The ability to simultaneously measure multiple secreted proteins and the corresponding gene expression levels from a single sample is valuable for comprehensive analysis. Bottlenecks to traditional immunoassays and gene expression assays include large sample consumption, time consuming experimental procedures, and complex data analysis.' ... More