ProcartaPlex™ Platinum Assay Buffer for Plasma Samples
Invitrogen™

ProcartaPlex™ Platinum Assay Buffer for Plasma Samples

ProcartaPlex Platinum Assay Buffer for Plasma Samples is used for dilution of plasma samples and gives spike- and dilution-recovery results in the range of 70 to 130%.
Have Questions?
Catalog NumberQuantity
EPXP-11112-00010 mL
Catalog number EPXP-11112-000
Price (INR)
9,614.00
Online offer
Ends: 31-Dec-2026
16,023.00
Save 6,409.00 (40%)
Each
Quantity:
10 mL
Price (INR)
9,614.00
Online offer
Ends: 31-Dec-2026
16,023.00
Save 6,409.00 (40%)
Each

ProcartaPlex Platinum Assay Buffer for Plasma Samples is used for dilution of plasma samples and gives spike- and dilution-recovery results in the range of 70–130%.

ProcartaPlex Platinum Assay Buffer for Plasma Samples is included in our Platinum ProcartaPlex Human Basic Kit for Plasma Samples (Cat.No. EPXP010-10421-901) and off-the-shelf Platinum panels. It is offered here as a stand-alone item.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Equipment)Luminex™ Instruments
Product LineProcartaPlex
Sample TypePlasma
Shipping ConditionWet Ice
Product TypeBuffer
Quantity10 mL
SpeciesHuman
Unit SizeEach
Contents & Storage
Store in refrigerator 2°C to 8°C

Frequently asked questions (FAQs)

What species can ProcartaPlex Platinum Assay Buffer for Plasma Samples be used with?

ProcartaPlex Platinum Assay Buffer for Plasma Samples is designed to be used with the ProcartaPlex Platinum assay format. Currently, this format has been validated for human samples only. Other species have not been tested.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

What kits can ProcartaPlex Platinum Assay Buffer for Plasma Samples be used with?

ProcartaPlex Platinum Assay Buffer for Plasma Samples is designed to be used with the ProcartaPlex Platinum assay format. It is used for dilution of plasma samples and gives spike- and dilution-recovery results in the range of 70-130%. ProcartaPlex Platinum Assay Buffer for Plasma Samples is included in our Platinum ProcartaPlex Human Basic Kit for Plasma Samples (Cat.No. EPXP010-10421-901) and off-the-shelf Platinum panels. It is also offered as a stand-alone item (Cat. No. EPXP-11112-000).

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Are the standalone ProcartaPlex assay buffers lot-specific such that they will only work with specific lots of other components?

No, the standalone ProcartaPlex assay buffers are not lot-specific.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can I get the catalog numbers for ProcartaPlex assay buffers and plates?

Here are the catalog numbers:

- EPX-11111-000: ProcartaPlex Universal Assay Buffer (1X) (10 mL) - It replaces all discontinued Procarta buffers (standard, assay, sample buffer); it is the same as the discontinued Procarta Cell Lysate Assay Buffer, 10 plate (Cat. No. PC0165), but only for one plate
- EPX-11110-000: ProcartaPlex Universal Assay Buffer (10X) (10 mL)
- EPXP-11112-000: ProcartaPlex Platinum Assay Buffer for Plasma Samples (1X, 10 mL)
- EPXP-11113-000: ProcartaPlex Platinum Assay Buffer for Serum Samples (1X, 10 mL)
- EPX-99999-000: ProcartaPlex Cell Lysis Buffer; It is the same as the discontinued Procarta Cell lysis Buffer (for cell lysate preparation) (Cat. No. PC6002)
- EPX-22222-000: ProcartaPlex Detection Antibody Diluent, Mouse (1X, 3 mL)
- EPX-33333-000: ProcartaPlex Detection Antibody Diluent, Human (1X, 3 mL)
- EPX-66666-000: ProcartaPlex Wash Buffer 10X (20 mL)
- EPX-66666-001: ProcartaPlex Wash Buffer 10X (200 mL)
- EPX-77777-000: ProcartaPlex Reading Buffer (1X, 40 mL)
- EPX-SAPE-000: ProcartaPlex Streptavidin-PE (1X, 5 mL) - 4.3 µg/mL (can range from 4.25-4.35 µg/mL)
- EPX-44444-000: ProcartaPlex 96 well Flat Bottom Plate, black (1 plate)
- EPX-88888-000: ProcartaPlex 96 well Filter Plate (1 plate)
- EPX-38488-000: ProcartaPlex 384 well Filter Plate (1 plate)

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.