Sensititre™ Equine EQUIN2F Vet AST Plate
Thermo Scientific™

Sensititre™ Equine EQUIN2F Vet AST Plate

Expand antimicrobial susceptibility testing for equine isolates using microbroth dilution ranges that meet CLSI veterinary recommendations
Have Questions?
Catalog NumberQuantity
EQUIN2F10/Pk.
Catalog number EQUIN2F
Price (INR)
-
Quantity:
10/Pk.
Meet the demand for rigorous equine antimicrobial susceptibility testing using the latest antibiotic and CLSI veterinary breakpoints with the Thermo Scientific™ Sensititre™ Equine EQUIN2F AST Plate. Accurate equine-specific MIC results on a single microbroth dilution plate, enable faster, more effective therapy decisions to support improved animal outcomes.

Demand for rigorous testing of animals and livestock with the latest antimicrobials and CLSI veterinary breakpoints has escalated due to emergent diseases and evolving multidrug resistance.

Perform accurate equine-specific susceptibility testing for reliable, consistent results and confident decisions using the Sensititre Equine EQUIN2F AST Plate.

  • Latest antimicrobials: ranges cover the CLSI breakpoints for amikacin, cefazolin, ceftiofur, enrofloxacin, clarithromycin, doxycycline, and tetracycline
  • Accurate MIC results: equine-specific microbroth dilution AST format using direct growth measurement provides accurate dosage information
  • Extended shelf life: room temperature storage eliminates inventory control concerns.
  • Reduced waste: individual packaging allows the laboratory to test one plate at a time with no waste.
  • Confidence in results: inclusive on-scale QC ranges provide immediate quality assurance of testing methodology.
  • Flexibility: automated or manual read options are available for ultimate flexibility.
  • Not all products are available for sale in all territories. Please inquire.
  • Sensititre™ products are now part of the Thermo Scientific brand.
Contact Thermo Fisher Scientific Microbiology Technical Support to add new breakpoint updates to your Thermo Scientific™ Sensititre™ SWIN™ Software
For Veterinary Use Only.
Specifications
0.5 McFarland StandardDemineralized water (part no. T3339)
Antibiotic CodeVarious
Antimicrobial AgentsVarious
Broth TypeMHB (part no. T3462)
DescriptionSensititre Equine EQUIN2F AST Plate
For Use With (Application)Gram negative and Gram postivie non-fastidious organisms from Equine isolates
Incubation Conditions34-36°C; non-CO2
Incubation Time18-24 hours
Quantity10/Pk.
Reading OptionsAutoread or manual
Specimen TypeEquine Isolates
Transfer Volume10µl
Volume Per Well50 μL
Product TypeAST Plate
SpeciesEquine
Target Organism ClassNon-fastidious, Gram-positive and Gram-negative Veterinary Isolates
Test TypeVeterinary AST, Minimum Inhibitory Concentration (MIC)
Unit SizeEach