Sensititre™ ESB1F
Thermo Scientific™
Sensititre™ ESB1F
Obtain accurate susceptibility results for Enterobacteriacea, as well as ESBL Confirmation, with the Thermo Scientific™ Sensititre™ Extended Spectrum Beta-lactamase Plate.
Have Questions?
Catalog NumberQuantity
ESB1F10/Pk.
Catalog number ESB1F
Price (INR)
-
Quantity:
10/Pk.
Obtain accurate susceptibility results for Enterobacteriacea, as well as ESBL Confirmation, with the Thermo Scientific™ Sensititre™ Extended Spectrum Beta-lactamase Plate. Extensive dilution ranges of confirmatory antimicrobics ensure full compliance with NCCLS recommendations, and a comprehensive selection of antimicrobials aid in selecting the most appropriate therapeutic treatment.
Obtain accurate susceptibility results for Enterobacteriacea, as well as ESBL Confirmation, with the Sensititre Extended Spectrum Beta-lactamase Plate.
  • Manual or automated read options provide flexible alternatives for fully automated or back-up test procedures
  • Long shelf life and room-temperature storage simplify inventory control and reduce expiry costs
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
0.5 McFarland StandardH2O (Part No. T3339)
AST TypeMIC
Antibiotic CodeVarious
Antimicrobial AgentsVarious
Broth TypeMHB (Part No. T3462)
DescriptionSensititre Extended Spectrum Beta-lactamase Plate
Incubation Conditions35°C O2
Incubation Time18 hr
Isolates Per Plate1
No. per Pack10
Packaging TypeBox
Quantity10/Pk.
Reading OptionsAutoread or manual
Transfer VolumeFor Non-Fastidious Gram Negative organisms, including suspected ESBL producers: 10μL, 30μL* (*Aid in detection of resistance mechanisms for gram negative organisms; see pack insert). For Proteus sp. (for research use only): 1μL
Volume Per Well50 μL
Product TypeAST Plate
Target Organism ClassNon-fastidious, Gram-negative Organisms
Test TypeMinimum Inhibitory Concentration (MIC), Breakpoint
Unit SizeEach