Sensititre™ EUMDR2F
Sensititre™ EUMDR2F
Sensititre™ EUMDR2F
Thermo Scientific™
Sensititre™ EUMDR2F
Confidently perform Antimicrobial Susceptibility Testing (AST) of clinically relevant non-fastidious Gram-negative isolates on a single comprehensive panel now containing Aztreonam / Avibactam, Cefepime / Enmetazobactam, and Sulbactam / Durlobactam.
Have Questions?
Catalog NumberTest Type
EUMDR2FMinimum Inhibitory Concentration (MIC)
Catalog number EUMDR2F
Price (INR)
-
Test Type:
Minimum Inhibitory Concentration (MIC)
Confidently perform Antimicrobial Susceptibility Testing (AST) of clinically relevant non-fastidious Gram-negative isolates on a single comprehensive AST panel now containing Aztreonam / Avibactam, Cefepime /Enmetazobactam, and Sulbactam / Durlobactam. Generate clinically impactful data, detect emerging resistance, and help guide timely and accurate antimicrobial therapy by utilising micro-broth dilution - the gold standard1 phenotypic method for AST.
This convenient panel meets EUCAST breakpoints for a broad range of relevant second line antimicrobials, including Ceftazidime / Avibactam, Colistin, Eravacycline, Imipenem / Relebactam, and Meropenem / Vaborbactam.
Key Features & Product Benefits
  •  Deliver consistent turnaround times to support earlier clinical decision-making and targeted therapy, particularly in critical infections such as sepsis.
  • Monitor susceptibility among critical Gram-negative isolates with full-range, accurate MIC values for a broad array of clinically relevant antimicrobics.
  • For ultimate flexibility automated or manual read options are available with one panel; compatible with all Sensititre™ instrumentation options - designed to be modular in order to meet the unique needs of individual lab space, workload, and budget requirements.
  • Eliminate inventory control concerns with extended shelf life (24-months), and room temperature storage.
  • Individual packaging allows testing of one sample at a time, with reduced waste.
Order now and transform your testing program with the latest AST panels.
Not all products are available for sale in all territories. Please inquire.
For In Vitro Diagnostic Use.  
1Jayol A, Nordmann P, Andre C, Poirel L, Dubois V. Evaluation of three broth microdilution systems to determine colistin susceptibility of Gram-negative bacilli. J Antimicrob Chemother. 2018; 73: 1272-1278. PubMed
Specifications
0.5 McFarland StandardWater (SKU: T3339)
Antimicrobial AgentsAmikacin, Aztreonam, Aztreonam / Avibactam Constant 4, Cefepime, Cefepime / Enmetazobactam Constant 8, Ceftazidime / Avibactam Constant 4, Ceftolozane / Tazobactam Constant 4, Colistin, Eravacycline, Imipenem / Relebactam Constant 4, Meropenem, Meropenem / Vaborbactam Constant 8, Negative Control, Piperacillin / Tazobactam Constant 4, Sulbactam / Durlobactam Constant 4, Tigecycline, Tobramycin
Broth TypeMueller-Hinton (SKU: T3462)
Incubation Conditions35°C, O2
Incubation Time18 hr.
Packaging TypeBox/10 panels
Specimen TypeNon-fastidious, Gram-negative
Volume Per Well50 μL
Product TypeAST Panel
SpeciesNon-fastidious Gram-negative Isolates
Target Organism ClassNon-fastidious Gram-negative Organisms
Test TypeMinimum Inhibitory Concentration (MIC)
Unit SizeEach
Contents & Storage
24 months
Frequently asked questions (FAQs)
What differentiates the Sensititre EUMDR2F panel from other Gram-negative AST panels?
The Sensititre EUMDR2F panel includes multiple antimicrobial combinations, such as aztreonam / avibactam, cefepime / enmetazobactam, and sulbactam / durlobactam, within a single broth microdilution format. This supports broader testing coverage for Gram-negative organisms.
When should I choose the Sensititre EUMDR2F panel for antimicrobial susceptibility testing?
The Sensititre EUMDR2F panel may be appropriate when testing non-fastidious Gram-negative isolates and when access to multiple antimicrobial combinations within a single MIC panel is needed. It is designed to support standardized broth microdilution testing in routine laboratory workflows.
Does the Sensititre EUMDR2F panel include cefepime / enmetazobactam and sulbactam / durlobactam?
Yes. The Sensititre EUMDR2F panel includes both cefepime / enmetazobactam and sulbactam / durlobactam on the same panel. This supports testing of multiple relevant Gram-negative antimicrobial combinations within a single MIC-based format.
Can the Sensititre EUMDR2F panel be used for aztreonam / avibactam susceptibility testing?
Yes. The Sensititre EUMDR2F panel includes aztreonam / avibactam for broth microdilution testing. This allows laboratories to generate MIC-based susceptibility data for this antimicrobial combination within a standardized AST workflow.
Is the Sensititre EUMDR2F panel suitable for testing non-fastidious Gram-negative isolates?
Yes. The Sensititre EUMDR2F panel is designed for clinically relevant non-fastidious Gram-negative isolates and uses broth microdilution to generate quantitative MIC results as part of the Sensititre antimicrobial susceptibility testing (AST) workflow. This supports standardized susceptibility testing for relevant organisms.