The convenient calibration kit contains xMAP® calibrators for MagPlex and MicroPlex classification channels and also includes the calibrator for theRead more
Have Questions?
Catalog Number
Quantity
F3DCALK25
25 reactions
Catalog number F3DCALK25
Price (INR)
-
Quantity:
25 reactions
The convenient calibration kit contains xMAP® calibrators for MagPlex and MicroPlex classification channels and also includes the calibrator for the reporter channel. Lot specific CD is included for importing target values. The kit is intended to be used with the off plate reagent area. Stripwells are included with each kit. Each kit will provide 25 calibrations.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineLuminex
Quantity25 reactions
Shipping ConditionWet Ice
For Use With (Equipment)FlexMAP 3D System
Product TypeCalibration Kit
Unit SizeEach
Contents & Storage
Contains 28 strip wells, FM3D Calibration Kit CD, Luminex Classification Calibrator Microspheres, MagPlex Classification Calibrator Microspheres, Luminex Reporter Calibrator Microspheres. Store at 2°C to 8°C.
Frequently asked questions (FAQs)
How long does it take to acquire ProcartaPlex assay data on the different Luminex instruments?
With everything working smoothly, here is the approximate read time for one plate on different Luminex instruments:
- FLEXMAP 3D Systems and INTELLIFLEX Systems will read 96 well plates in ~20 min, and 384 well plates in ~75 min.
- Luminex 100/200 Systems will read 96 well plates in ~40 min.
- MAGPIX System will read 96 well plates in ~50-60 min. Please note that these are estimates for the read times of the plates only, and do not include time for calibration or set-up of the instrument.