Orion™ ISE Calibration Standards
Thermo Scientific™

Orion™ ISE Calibration Standards

Take reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Calibration Standards. For use with ion selective electrodes from ammonia to water hardness.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberAnalyte or Component NamesConcentration
F40905Fluoride5 ppm with TISAB II
951006Ammonia0.1M
951207Ammonia100 ppm
943506Bromide0.1M
922006Calcium0.1 M
950207Carbon Dioxide1000 ppm
941706Chloride and Sodium0.1 M
941708Chloride1000 ppm
941707Chloride100 ppm
977007Residual Chlorine100 ppm
942906Cupric0.1 M
940906Fluoride0.1 M
940907Fluoride100 ppm
040908Fluoride10 ppm with TISAB II
040907Fluoride2 ppm with TISAB II
040906Fluoride1 ppm with TISAB II
F40904Fluoride0.5 ppm with TISAB II
940916Fluoride1 ppm with TISAB II and 10 ppm with TISAB II
945306Iodide0.1 M
948206Lead0.1 M
920706Nitrate0.1 M KNO3
920707Nitrate1000 ppm
930707Nitrate100 ppm
921906Potassium0.1 M
941107Sodium100 ppm
941105Sodium10 ppm
923206Water Hardness100 ppm
Catalog number F40905
Price (INR)
-
Analyte or Component Names:
Fluoride
Concentration:
5 ppm with TISAB II

Take accurate, reproducible ion concentration measurements by calibrating your Ion Selective Electrode with high-quality Thermo Scientific™ Orion™ ISE Calibration Standards. Use these standards with electrodes including ammonia, chloride, fluoride, nitrate, potassium, silver, sodium and sulfide. Orion calibration standards are NIST traceable and manufactured under ISO 9001 quality standards.

Use the Orion ISE calibration standards with our extensive line of ion selective electrodes (ISEs) for ammonia, ammonium, bromide, cadmium, calcium, carbon dioxide, chloride, chlorine, copper, cyanide, fluoride, iodide, lead, nitrate, potassium, silver, sodium and sulfide analysis. Whether you are in the lab or in the field, you can be confident that Orion solutions will deliver accurate and repeatable measurements. Orion solutions are produced in controlled batches using ultrapure water and undergo rigorous quality testing throughout the process to prevent contamination. Our strict quality procedures verify correct bottle labeling and volume, so each batch meets specific lot release criteria. Bottles are sealed and torque-tested to ensure bottle caps are secure and solutions remain free of contamination. Rigorous lifetime testing ensures sealed Orion solutions remain within published specifications for their entire shelf life and do not degrade over time.

 

* Orion calibration standards are NIST traceable.

 

* Orion calibration standards are manufactured under ISO 9001 quality standards.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Analyte or Component NamesFluoride
Certificate of AnalysisYes
ColorOrange
Concentration or Composition NotesC of A Acceptance Range: 2.50 ppm +/- 0.05 ppm Fluoride (50:50 Premix of Standard to TISAB II)
DescriptionFluoride Standard
For Use With (Equipment)Fluoride Ion Selective Electrode
Packaging1 Bottle
Physical FormLiquid
Product LineFluoride Ion Selective Electrodes Including 9609BNWP, 9409BN, 9409SC
Quantity475 mL
Solution TypeCalibration Standard
Traceability to NISTYes
TypeCalibration Standard
Concentration5 ppm with TISAB II
Unit SizeEach