FluoSpheres™ Sulfate-Modified Microspheres, 0.02–4.0 μm
Invitrogen™

FluoSpheres™ Sulfate-Modified Microspheres, 0.02–4.0 μm

Perform flow cytometry, microscopy, HTS, and HCS with our FluoSpheres Sulfate-Modified Microspheres, featuring different bead diameters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Color
F88521 μmYellow-Green
F8851
also known as F-8851
1 μmRed
F88584 μmRed
F8845
also known as F-8845
0.02 μmYellow-Green
F88480.20 μmYellow-Green
F88532 μmYellow-Green
F88594 μmYellow-Green
Catalog number F8852
Price (INR)
-
Diameter (Metric):
1 μm
Color:
Yellow-Green
Easily perform flow cytometry, microscopy, HTS, HCS, and other laboratory applications using our extensive selection of FluoSpheres Sulfate-Modified Microspheres. These microsphere beads can be used in passive adsorption or active, covalent coupling of proteins, nucleic acids, and biomolecules for particle capture applications.

FluoSpheres Sulfate-Modified Microspheres are loaded with our proprietary fluorescent dyes, making them the brightest microspheres available. Because the fluorescent dye molecules are contained inside each polystyrene microsphere instead of only on the surface, the dye molecules are shielded from many of the environmental effects that cause quenching or photobleaching of exposed fluorophores. The stability, uniformity, and reproducibility of FluoSpheres microspheres, as well as their extensive color selection, make them the preferred tools for research and diagnostic assays that use fluorescence.

FluoSpheres Sulfate-Modified Microspheres are relatively hydrophobic particles that will passively and nearly irreversibly adsorb almost any protein, including albumin, IgG, avidin, and streptavidin.

Key applications of FluoSpheres Sulfate-Modified Microspheres
• Instrument calibration (flow cytometry, microscopy, HTS, HCS)
• Flow testing (microfluidics, blood flow, water flow, and air flow)
• Cell biology tracers (cell differentiation and cell tracing)
• Immunoassays (agglutination tests, ELISA, particle capture, and contrast reagents)

FluoSpheres sulfate-modified microsphere specifications for F8845, F8848, F8852, F8853, and F8859:
• Label (excitation/emission): yellow-green (505/515)
• Nominal bead diameters: 0.02, 0.2, 1.0, 2.0, and 4.0 μm
• Coupling surface: sulfate
• Solids: 2%

FluoSpheres sulfate-modified microsphere specifications for F8851 and F8858:
• Red (580/605)
• Nominal bead diameters: 1.0, 4.0 μm
• Coupling surface: sulfate
• Solids: 2%

FluoSpheres fluorescent microspheres are potentially more sensitive than colorimetric methods in most, if not all, major microsphere-based diagnostic test systems presently in use, including microsphere-agglutination tests, filter separation tests, particle-capture ELISA methods, and two-particle sandwich techniques.

FluoSphere Sulfate-Modified Microspheres (also called latex beads or latex particles) are manufactured using high-quality, ultra-clean polystyrene and are loaded with a variety of our proprietary dyes to create intensely fluorescent beads that typically show little or no photobleaching, even when excited with the intense illumination required for fluorescence microscopy. Every possible precaution is taken throughout the manufacturing process to ensure that the microparticles are kept free of contaminating agents. The final product is sold as a suspension in ultrapure water, in most cases containing 2 mM azide or 0.02% thimerosal as a preservative.

Other surface modifications are available to facilitate the coupling of various molecules and proteins to the surface of the bead, including amine- and carboxylate-modified FluoSphere microspheres.

Choices for unstained microspheres
We also offer hundreds of choices for ultra-clean surfactant-free microspheres for research and commercial applications.

Custom microspheres
We prepare custom microspheres upon request. For example, FluoSpheres Sulfate-Modified microsphere beads can be prepared with intensities that are lower than those of our regular selection, a desirable feature in some multicolor applications. Our custom conjugation service is efficient and confidential, and we guarantee the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineFLUOSPHERES
Quantity10 mL
Shipping ConditionRoom Temperature
Surface ModificationSulfate
Volume (Metric)10 mL
ColorYellow-Green
Diameter (Metric)1 μm
For Use With (Application)Cell Analysis
MaterialPolystyrene
Product TypeMicrosphere
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.

Frequently asked questions (FAQs)

I have some FluoSpheres polystyrene microspheres, with 20 nm diameter. They are aggregating a lot. What can I do about it?

The smaller the microspheres, the greater the propensity to aggregate. But the aggregation is not irreversible. Sonicate in a bath sonicator or vortex to disperse, just prior to use. You can also add a small concentration of Tween-20 or Triton X-100 (unless you are using them in a live-cell system).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I sonicated my 2.0 µm carboxylate-modified microspheres, as recommended, but saw foaming (bubbles) on top of the solution. Should I be concerned?

Use of a bath sonicator is recommended to help break up any aggregated microspheres. The foaming is from Tween-20, which is in the stock solution to help prevent aggregation. It is normal and expected to see bubbles from this. Do not use a probe sonicator, which would cause damage to the microspheres (as well as much more bubbling).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What is the warranty for FluoSpheres microspheres?

The warranty period for FluoSpheres microspheres is 1-year from the date of shipment.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

After washing and centrifugation, there was only a very small pellet left of my microsphere beads and the solution was transparent. Why is this?

Centrifugation is not an effective way to collect smaller microspheres; many particles remain in the solution even if you can visualize a small pellet. For beads less than 1 µm in diameter, we recommend washing by either:

Cross-flow filtration, as these particles have a very high compression modulus and can withstand high g-forces without risk of harm or dialysis with a 500 kDa MWCO
Note: Microspheres greater than 1 µm in diameter can be centrifuged at 1,300 rpm.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I've had my microspheres for over a year, and I'm wondering if they're still good to use. What are some good ways to check their functionality?

Bacterial contamination is the most common cause of microspheres becoming unusable. Many of our particles are supplied with a low level of sodium azide to prevent bacterial contamination, but sometimes this can still occur. Bacterial contamination is best assessed by plating on appropriate growth medium and checking the plates after 72 hr.

Find additional tips, troubleshooting help, and resources within ourMicrospheres Support Center.

Citations & References (6)

Citations & References
Abstract
CR1-mediated ATP Release by Human Red Blood Cells Promotes CR1 Clustering and Modulates the Immune Transfer Process.
Authors:Melhorn MI, Brodsky AS, Estanislau J, Khoory JA, Illigens B, Hamachi I, Kurishita Y, Fraser AD, Nicholson-Weller A, Dolmatova E, Duffy HS, Ghiran IC,
Journal:
PubMed ID:24022490
'Humans and other higher primates are unique among mammals in using complement receptor 1 (CR1, CD35) on red blood cells (RBC) to ligate complement-tagged inflammatory particles (immune complexes, apoptotic/necrotic debris, and microbes) in the circulation for quiet transport to the sinusoids of spleen and liver where resident macrophages remove the ... More
A simple method for monitoring changes in cell height using fluorescent microbeads and an Ussing-type chamber for the inverted microscope.
Authors:Crowe WE, Wills NK
Journal:Pflugers Arch
PubMed ID:1745609
'In this study, we report two developments for studies of ion transport in cultured epithelial cells. First, a convenient method is presented for measuring apparent cell height using fluorescent microbeads as high-contrast landmarks of the apical and basal cell surfaces. The apparent cell height is then used as an indicator ... More
Swelling-activated cation-selective channels in A6 epithelia are permeable to large cations.
Authors:Li J, De Smet P, Jans D, Simaels J, Van Driessche W
Journal:Am J Physiol
PubMed ID:9688589
'Effects of basolateral monovalent cation replacements (Na+ by Li+, K+, Cs+, methylammonium, and guanidinium) on permeability to 86Rb of volume-sensitive cation channels (VSCC) in the basolateral membrane and on regulatory volume decrease (RVD), elicited by a hyposmotic shock, were studied in A6 epithelia in the absence of apical Na+ uptake. ... More
Transitional states of acrosomal exocytosis and proteolytic processing of the acrosomal matrix in guinea pig sperm.
Authors:Kim KS, Foster JA, Kvasnicka KW, Gerton GL,
Journal:Mol Reprod Dev
PubMed ID:21919109
In this study, we adapted a FluoSphere bead-binding assay to study the exposure and release of guinea pig sperm acrosomal components during the course of capacitation and acrosomal exocytosis. Prior to capacitation or the initiation of exocytosis, acrosomal proteins were not accessible to FluoSpheres coated with antibodies against two acrosomal ... More
An automatic monitoring system for epithelial cell height.
Authors:Van Driessche W, De Smet P, Raskin G
Journal:Pflugers Arch
PubMed ID:8272373
This paper describes an automatic method to measure cell height (h) of epithelia grown as monolayers on transparent filter supports. Tissues are mounted in an Ussing-type chamber enabling solution exchange on both sides. The apical and basal side of the epithelial cells are marked with fluorescent beads. The image of ... More