Product Image
Thermo Scientific™
FDP80 Oil Vacuum Pumps
Perform standard freeze-drying applications with specifically designed Thermo Scientific™ FDP80 Oil Vacuum Pumps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVoltage
FDP80-115115 V
FDP80-230230 V
Catalog number FDP80-115
Price (INR)
-
Voltage:
115 V
Perform standard freeze-drying applications with specifically designed Thermo Scientific™ FDP80 Oil Vacuum Pumps.
Features:
  • 76L/min. (at 60Hz) throughput
  • 1.5mtorr ultimate vacuum level
  • Gas Ballast Oil Recovery Kit (GBORK) allows the user to apply full gas ballast to keep the oil clean without losing oil
  • Oil mist filter filters the exhaust to keep environmental air clean
Ordering Information:
For applications involving aggressive solvents that might damage ordinary pumps, we recommend the LyoPump VLP195FD which also includes an active oil filtration system (VPOF110) that dramatically extends the life of the pump.
Includes:
GBORK (Gas Ballast Oil Return Kit) and Oil Mist Filter (EMF-10).
Specifications
ApplicationsStandard Freeze Drying
Electrical Requirements115 V 60 Hz
Frequency60 Hz
Product LineThermo Scientific
Flow Rate76 L/min
Max. Vacuum1.5 mtorr
Pump TypeOil Vacuum Pump
Voltage115 V
Unit SizeEach