Forma™ Flammable Storage Freezer, -20°C; 23 cu. ft., 115V
Thermo Scientific™

Forma™ Flammable Storage Freezer, -20°C; 23 cu. ft., 115V

Protect flammable materials, solvents and intermediates using Thermo Scientific™ Forma™ Flammable Materials Freezers.
Have Questions?
Catalog NumberCapacity (English)Capacity (Metric)Voltage
FFMS2320A23.3 cu. ft.659 L115 V
Catalog number FFMS2320A
Price (INR)
-
Capacity (English):
23.3 cu. ft.
Capacity (Metric):
659 L
Voltage:
115 V

Protect flammable materials, solvents and intermediates using high-performance Thermo Scientific™ Forma™ Flammable Materials Freezers. Designed to meet stringent U.S. Pharmacopeia standards for drug storage and  NFPA safety provisions for storing flammable liquids.† These freezers are designed to maintain a safe environment while keeping valuable samples within a tight temperature range. 


Each is listed by UL as a special purpose freezer intended for flammable material storage in accordance with NFPA 45, 70 and 99.† 45, 70 and 99.†

Key Features:
  • Easy to use intuitive digital display featuring microprocessor control
  • Lockable doors, audio and visual alarms including door ajar alarms for security and peace of mind
  • Convenient access port and casters
  • Outstanding construction featuring painted, high-quality steel exteriors with durable finish
  • Ignition-free interior cabinet design for the safe storage of flammable materials
  • Meets safety standards in accordance with UL, NFPA† and OSHA† for regulatory compliance
  • Optional, built-in, inkless, seven-day graphic chart recorder
  • Optional, barrier-protected 4-20 milliamp output for external monitoring
  • Manual defrost

Ordering Information:

Temperature chart recorders, and 4-20 milliamp are available separately.

Note:

† Products complying with the applicable requirements of NFPA 70, “National Electric Code”, NPFA 45, “Fire Protection for Laboratories Using Chemicals,” and NFPA 99 “Health Care Facilities” are considered to meet the OSHA guidelines for these types of facilities. These units have no internal electrical components that could trigger an explosion or fire of flammable materials inside the unit. Flammable material storage units are not for use in classified hazardous locations which require the use of explosion-proof units.

Specifications
Temperature Range-20°C
Certifications/ComplianceCE, cUL, UL
Capacity (English)23.3 cu. ft.
Capacity (Metric)659 L
Electrical Requirements115 V 60 Hz
Voltage115 V
Frequency60 Hz
Amperage13.5 A
Dimensions (D x W x H) Interior29 x 24 x 58 in. (73.7 x 61 x 147.3 cm)
RefrigerantCFC-free
Breaker20A
Depth (English) Interior29 in.
Depth (Metric) Interior73.7 cm
Height (English) Exterior78.6 in.
Height (English) Interior58 in.
Height (Metric) Exterior200 cm
Height (Metric) Interior147.3 cm
Width (English) Interior24 in.
Width (Metric) Interior61 cm
DefrostManual
Width (English) Exterior28 in.
Width (Metric) Exterior72 cm
Depth (English) Exterior37 in.
Depth (Metric) Exterior94 cm
Door StyleLockable, Audio and Visual Alarms Including
Cabinet MaterialScratch-resistant Painted Steel
InsulationCFC-free
MaterialSteel
Monitoring OptionsExternal
No. of Doors1
Shelves4
Unit SizeEach