E-Gel™ Power Snap Electrophoresis Device Starter Kit, SYBR Safe 1%
Invitrogen™
E-Gel™ Power Snap Electrophoresis Device Starter Kit, SYBR Safe 1%
Provides everything needed to quickly start using precast 1% E-Gel agarose gels with SYBR Safe on the device.
Have Questions?
Catalog NumberQuantity
G8121ST1 Kit
Catalog number G8121ST
Price (INR)
192,218.00
Each
Quantity:
1 Kit
Price (INR)
192,218.00
Each
The Invitrogen E-Gel Power Snap Electrophoresis Device Starter Kit, SYBR Safe 1%, provides everything you need to quickly start using precast 1% E-Gel agarose gels with SYBR Safe on the device.
The kit includes:
  • 10 E-Gel agarose gels with SYBR Safe, 1% (Cat. No. A42100)
  • E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
  • E-Gel 1 Kb Plus DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488090)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 Kit
Shipping ConditionRoom Temperature
Max. Voltage240 VAC
Product LineE-Gel
TypeElectrophoresis System Starter Kit
Unit SizeEach
Contents & Storage
• 1 electrophoresis device
• 10 E-Gel agarose gels with SYBR Safe, 1%
• 1 vial E-Gel 1 kb Plus DNA Ladder
• 1 power adapter
• 1 pair of viewing glasses
Store at room temperature.