E-Gel™ Power Snap Electrophoresis Device Starter Kit, EX DC 2%
Invitrogen™
E-Gel™ Power Snap Electrophoresis Device Starter Kit, EX DC 2%
The Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, EX DC 2%, provides everything you need to quickly start using precast E-Gel™ EX Double Comb 2% agarose gels on the device.
Have Questions?
Catalog NumberQuantity
G8132ST1 Kit
Catalog number G8132ST
Price (INR)
-
Quantity:
1 Kit
The Invitrogen E-Gel Power Snap Electrophoresis Device Starter Kit, EX DC 2%, provides everything you need to quickly start using precast E-Gel EX Double Comb 2% agarose gels on the device.
The kit includes:
  • Ten E-Gel EX Double Comb Agarose Gels, 2% (Cat. No. A42346)
  • E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
  • E-Gel 50-bp DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488099)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 Kit
TypeElectrophoresis System Starter Kit
Unit SizeEach
Contents & Storage
• Ten E-Gel EX Double Comb Agarose Gels, 2% (Cat. No. A42346)
• E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
• E-Gel 50-bp DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488099)
Store at room temperature.
Frequently asked questions (FAQs)
I am not seeing any current when I try to run my E-Gel EX agarose gels. Why is this?
Here are some common reasons why your gel is not running properly:
- Copper contacts in the base are damaged due to improper use. Make sure the copper contacts in the base are intact.
- An expired or defective gel cassette was used.
- The E-Gel EX cassette was not inserted properly into a base.
- An incorrect adaptor was used.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.
What is the special feature of E-Gel EX agarose gels?
E-Gel EX agarose gels separate DNA faster, offer enhanced sensitivity, and provide added flexibility. The stain within the EX gels is proprietary (not based on SYBR technology), though it has the same spectral properties as SYBR stains. The E-Gel EX gels have a sensitivity that is 5X greater than gels using ethidium bromide.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.
I have an E-Gel Simple Runner Electrophoresis Device in my lab, are the new 11-well and 22-well E-Gel agarose gels compatible with it?
Due to the differences in dimensions, the new E-Gel 11-well and 22-well agarose gel formats are not compatible with the E-Gel Simple Runner Electrophoresis Device.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.
Which DNA ladders do you recommend using with E-Gel EX agarose gels and E-Gel agarose gels with SYBR Safe stain?
To select the DNA ladder that yields the best resolution for your specific E-Gel, please refer to the table on page 45 of the E-Gel Power Snap Electrophoresis System user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0017050_egel_powersnapsystem_UG.pdf).
Can I use double comb E-Gel EX agarose gels or E-Gel agarose gels with SYBR Safe stain for RNA analysis?
E-Gel EX Double Comb agarose gels and E-Gel Double Comb agarose gels with SYBR Safe stain have not been tested for use in RNA applications. We recommend using E-Gel EX Single Comb agarose gels with 11 wells for RNA analysis.