TRANSiPORT Human OATP1A2 (G) SLC Transporter Cells are HEK293 cells that transiently overexpress the OATP2B1 solute carrier (SLC) transport protein.Read more
Have Questions?
Catalog Number
Quantity
GM1116G
8–10M cells
Catalog number GM1116G
Price (INR)
72,744.00
Online offer
Ends: 31-Mar-2026
121,240.00
Save 48,496.00 (40%)
Each
Quantity:
8–10M cells
Price (INR)
72,744.00
Online offer
Ends: 31-Mar-2026
121,240.00
Save 48,496.00 (40%)
Each
TRANSiPORT Human OATP1A2 (G) SLC Transporter Cells are HEK293 cells that transiently overexpress the OATP2B1 solute carrier (SLC) transport protein. Solute carriers are a super-family of membrane transporters that can affect pharmacokinetics and drug exposure by governing the transport of solutes in and out of cells. The OATP1A2 SLC transporter protein is found in hepatocytes.
• Assess potential for transporter-mediated drug metabolism • Easy-to-use format • Obtain high-quality results with a large signal-to-noise ratio
Choice of measurement system Cell-based assays can be performed using radioisotope-labeled compounds, fluorescence-labeled compounds, or non-labeled compounds. The amount of substrate transported into the cells can be measured directly using a liquid scintillation counter, fluorescence plate reader, or LC-MS/MS, thereby allowing direct evaluation of SLC transporter activity.
Assay reliability Mock OATP1A2 (G) SLC Transporter Cells (Cat. No. GM1017) that do not overexpress the OATP1A2 SLC transporter are available for use as a negative experimental control. Some compounds may demonstrate high background levels of transport due to the presence of endogenous transporters or non-specific binding.
Rapid results The convenient product format enables data generation in two days from thawing of the cells to final results.
Specifications
No. of Cells8-10 x 106 cells
Product TypeSLC Transporter Cell
Quantity8–10M cells
Cell Line293 (HEK)
Culture TypeMammalian Cell Culture
SpeciesHuman
Unit SizeEach
Contents & Storage
Store in liquid nitrogen vapor phase.
Frequently asked questions (FAQs)
What are TRANSiPORT human solute carrier (SLC) transporter cells?
TRANSiPORT human SLC transporter cells are HEK293 cells that transiently overexpress a solute carrier (SLC) transporter protein. Solute carriers are one of two superfamilies of membrane transporters that can affect pharmacokinetics and drug exposure by governing the transport of solutes into and out of cells. Membrane transporters are found in hepatic cells, kidney proximal tubule cells, intestinal cells, and brain cells.
Is there a protocol available for the TRANSiPORT solute carrier (SLC) transporter cells?
Yes, the protocol is available here (https://www.genomembrane.com/information).
What should I use as a control with TRANSiPORT solute carrier (SLC) transporter cells?
Mock HEK293 cells (Cat. No. GM1001) that do not overexpress the SLC transporter are available for use as a negative experimental control. Some compounds may demonstrate high background levels of SLC transport due to the presence of endogenous transporters or non-specific binding.
With TRANSiPORT solute carrier (SLC) transporter cells, what should my choice of measurement be for SLC transporter activity?
Cell-based assays can be performed using radioisotope-labeled compounds, fluorescence-labeled compounds, or non-labeled compounds. The amount of substrate transported into the cells can be measured directly using a liquid scintillation counter, fluorescence plate reader, or LC-MS/MS, thereby allowing direct evaluation of SLC transporter activity.
How many cells can I expect in one vial of TRANSiPORT solute carrier (SLC) transporter cells?
One vial contains 8-12 million cells with high viability, designed to seed one 24-well plate for single use.