Sensititre™ Gram Negative GNX2F AST Plate
Sensititre™ Gram Negative GNX2F AST Plate
Sensititre™ Gram Negative GNX2F AST Plate
Thermo Scientific™
Sensititre™ Gram Negative GNX2F AST Plate
Perform susceptibility testing of Gram-negative isolates on a single platform including colistin and Polymixin B with this research use only plate.
Have Questions?
Catalog NumberQuantity
GNX2F10/Pk.
Catalog number GNX2F
Price (INR)
-
Quantity:
10/Pk.
Perform susceptibility testing of non-fastidious Gram-negative isolates on a single platform with Thermo Scientific™ Sensititre™ Gram Negative GNX2F AST Plate, designed for research use only.
Decrease costs associated with multiple test methods with 21 antimicrobial compounds on a single plate, including colistin and polymixin B.
Test most bacteria, including challenging resistant organisms, on a single platform with Sensititre Gram Negative GNX2F AST Plate. 
  • Automated or manual read options are available for ultimate flexibility
  • Extended shelf life and room-temperature storage simplify inventory control
  • Individual packaging allows laboratory to test one plate at a time with no waste
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
0.5 McFarland StandardH2O (Part No. T3339)
AST TypeMIC
Antibiotic CodeVarious
Antimicrobial AgentsVarious
Broth TypeMHB (Part No. T3462)
DescriptionSensititre Gram Negative Research Use Only MIC Plate for CRE Testing
Incubation Conditions35°C O2
Incubation Time18 hr
Isolates Per Plate1
No. per Pack10
Packaging TypeBox
Quantity10/Pk.
Reading OptionsAutoread or manual
Transfer VolumeFor Enterobacteriaceae and Non-Enterobacteriaceae: 10μL, 30μL* (*Aid in detection of resistance mechanisms for gram negative organisms; see pack insert). For Proteus sp.: 1μL
Volume Per Well50 μL
Product TypeAST Plate
Target Organism ClassNon-fastidious, Gram-negative Organisms
Test TypeMinimum Inhibitory Concentration (MIC), Carbapenem-resistant Enterobacterales (CRE)
Unit SizeEach