Ion ReproSeq™ PGT-A GX5 Kit
Ion Torrent™

Ion ReproSeq™ PGT-A GX5 Kit

Experience the convenience of all-in-one template preparation and sequencing, streamlining your PGT-A workflow with load-and-go functionality and user-friendly software. ForRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Samples
GX34903384 Samples
GX34902192 Samples
Catalog number GX34903
Price (INR)
5,716,548.00
Each
No. of Samples:
384 Samples
Price (INR)
5,716,548.00
Each

Experience the convenience of all-in-one template preparation and sequencing, streamlining your PGT-A workflow with load-and-go functionality and user-friendly software. For researchers seeking a comprehensive next-generation sequencing (NGS) workflow for aneuploidy analysis, the Ion ReproSeq PGT-A Kit for the Genexus Integrated System provides all the necessary reagents and materials. This kit enables a rapid, cost-effective, and end-to-end workflow for the detection of aneuploidies across all 24 chromosomes, including autosomes (22) and the X and Y chromosomes. It is an ideal solution for research labs focusing on reliable assessment of pre-implantation embryo samples.

With the Ion ReproSeq PGT-A Kit for the Genexus System, you can achieve accurate detection of whole chromosome and chromosome arm copy number events. This kit empowers you to identify copy number events as small as 10 Mbp. By incorporating Genexus 6.8 software analysis, you can enhance data interpretation with advanced functionalities such as mosaicism detection, gender masking, and improved data plotting.

Cost-effective NGS workflow with sample throughput flexibility
Achieve a cost-effective NGS workflow with flexible sample throughput using the Ion ReproSeq PGT-A Kit for the Genexus System. The 192/384 sample kit comes with two GX5 chips, allows 24/48 samples per lane, and the three unused lanes enable additional runs, spreading the 96/192 samples per chip requirement over two weeks. Alternatively, run all four lanes simultaneously for a throughput of 96/192 samples per run. The innovative GX5 chip design enhances scalability, cost efficiencies, and the ability to adapt to sample fluctuations and increased volume.

Rapid turnaround for time-sensitive samples
The Genexus System simplifies templating, sequencing, analyzing, and reporting for PGT-A samples. With integrated templating, sequencing, and Genexus software, it automates these steps effortlessly, tracking sample details and delivering accurate results. This automation reduces the need for manual intervention, generates reliable data, and produces rapid reports. From single or multiple cells from a pre-implantation embryo, you can obtain analyzed data in under 11 hours for up to 48 samples.

Notes
• This kit is only compatible with the Genexus System.
 

For Research Use Only.
Specifications
No. of Samples384 Samples
Quantity384 samples
FormatKit
Sequencing TypeNext Generation Sequencing
Unit SizeEach
Contents & Storage
• Ion SingleSeq Reagents, –15 to –25°C
• Ion SingleSeq Barcode Plate Sets, –15 to –25°C
• Genexus Cartridge, –10 to –30°C
• Genexus Bottle 2, 15–30°C
• Genexus Bottle 1 and 3, 15–30°C
• Genexus Strip 3-GX5, 2–8°C
• Genexus Strip 4, –10 to –30°C
• Genexus Coupler and GX5 Chip, 15–30°C

Frequently asked questions (FAQs)

With the Ion ReproSeq PGT-A GX5 Kit, what is the turnaround time (sample to test results) for PGT-A + SNP QC samples?

The turnaround time for PGT-A + SNP QC samples is estimated to be 21 hr.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

With the Ion ReproSeq PGT-A GX5 Kit, what is the turnaround time (sample to test results) for SNP QC-only samples?

The turnaround time for SNP QC-only samples is estimated to be 18 hr (48 samples).

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

With the Ion ReproSeq PGT-A GX5 Kit, what is the turnaround time (sample to test results) for PGT-A-only samples?

The turnaround time for PGT-A-only samples is estimated to be 10 hr 30 min (48 samples) and 19 hr 45 min (192 samples).

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What version of Genexus Software is needed to run the Ion ReproSeq PGT-A GX5 Kit?

Genexus Software version 6.8.0 is needed to run the Ion ReproSeq PGT-A GX5 Kit.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

With the Ion ReproSeq PGT-A GX5 Kit, how many reads per sample can I expect for 48 libraries/lane?

48 libraries per lane yields >200,000 reads for most libraries for the Ion ReproSeq PGT-A GX5 Kit.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.