1-Heptane Sulfonic Acid, Sodium Salt, Ion Pair Chromatography, Fisher Chemical™
1-Heptane Sulfonic Acid, Sodium Salt, Ion Pair Chromatography, Fisher Chemical™
1-Heptane Sulfonic Acid, Sodium Salt, Ion Pair Chromatography, Fisher Chemical™
Fisher Chemicals

1-Heptane Sulfonic Acid, Sodium Salt, Ion Pair Chromatography, Fisher Chemical™

1-Heptane Sulfonic Acid, Sodium Salt, Ion Pair Chromatography, Fisher Chemical, Quantity: 100g, Packaging: Plastic powder jar, CAS: 22767-50-6, White, Melting Point: 300 deg.C, Molecular Formula: C7H15NaO3S, Molecular Weight: 202.244
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic powder jar
Catalog number H016846
also known as H/0168/46
HSN 98020000
Price (INR)
7,031.00
Each
Quantity:
25 g
Packaging:
Plastic powder jar
Request bulk or custom format
Price (INR)
7,031.00
Each
Specifications
Melting Point300°C
CAS22767-50-6
ColorWhite
Physical FormPowder/Solid
Quantity25 g
View more
1-Heptane Sulfonic Acid, Sodium Salt, Ion Pair Chromatography, Fisher Chemical, Quantity: 100g, Packaging: Plastic powder jar, CAS: 22767-50-6, White, Melting Point: 300 deg.C, Molecular Formula: C7H15NaO3S, Molecular Weight: 202.244