Barnstead™ RO Distribution Pumps
Barnstead™ RO Distribution Pumps
Actual product may vary
Thermo Scientific™
Barnstead™ RO Distribution Pumps
Depend on the heavy-duty, split-phase induction motor in Thermo Scientific™ Barnstead™ RO Distribution Pumps, for use with pure water storage tanks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHorsepowerPhaseElectrical Requirements
H11221/2hp1115/230 V 50/60 Hz
H11121/3hp1115/230 V 50/60 Hz
H11321hp1115/230 V 50/60 Hz
H11421/2-1hp3230/460 V 50/60 Hz
Catalog number H1122
Price (INR)
-
Request A Quote
Horsepower:
1/2hp
Phase:
1
Electrical Requirements:
115/230 V 50/60 Hz
Depend on the heavy-duty, split-phase induction motor in Thermo Scientific™ Barnstead™ RO (Reverse Osmosis) Distribution Pumps. Distribution pumps are used with purified water storage tanks when it is necessary to keep the pure water under pressure for delivery to locations above tank level or at greater than gravity flow pressure.
Includes:
  • Plastic pump bypass with restrictive orifice
  • Low-level cutoff switch
Specifications
Electrical Requirements115/230 V 50/60 Hz
Frequency50/60 Hz
Horsepower1/2hp
Phase1
Voltage115/230 V
For Use With (Equipment)Purified Water Storage Tanks
Unit SizeEach