HybriWell™ Hybridization Sealing System, 40 mm x 21 mm chamber, 0.15 mm deep
Invitrogen™

HybriWell™ Hybridization Sealing System, 40 mm x 21 mm chamber, 0.15 mm deep

The HybriWell hybridization sealing system is a coverslip—seal combination that forms chambers optimized for in situ hybridization work. Each disposableRead more
Have Questions?
Catalog NumberQuantity
H182101 set
Catalog number H18210
Price (INR)
36,544.00
線上優惠
Ends: 30-Sep-2026
48,724.83
Save 12,180.83 (25%)
Each
Quantity:
1 set
Price (INR)
36,544.00
線上優惠
Ends: 30-Sep-2026
48,724.83
Save 12,180.83 (25%)
Each
The HybriWell hybridization sealing system is a coverslip—seal combination that forms chambers optimized for in situ hybridization work. Each disposable incubation chamber seals securely to a microscope slide, providing a limited-volume chamber for reagent addition. Solutions are easily added through dual-access ports without disrupting the specimen. The hydrophobic coverslips are nuclease free and will not trap or bind probes, allowing uniform distribution of the reagent over the specimen. The HybriWell sealing system also includes a quick-seal tool to secure the hydrophobic cover to the microscope slide, as well as nuclease-free adhesive seal tabs to cover the access ports (seal tabs are also available separately).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Depth (Metric)0.15 mm
Dimensions40 x 21 mm
Quantity1 set
Shipping ConditionRoom Temperature
Product LineHybriWell
TypeHybridization Sealing System
Unit SizeEach
Contents & Storage
Store at room temperature.